Skip to Content
Merck
All Photos(2)

Key Documents

L7533

Sigma-Aldrich

LB Broth with agar (Lennox)

EZMix powder microbial growth medium

Synonym(s):

LB broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.85
Pricing and availability is not currently available.

grade

for molecular biology

Quality Level

sterility

non-sterile

form

powder

composition

Agar, 15 g/L
NaCl, 5 g/L
Tryptone, 10 g/L
Yeast Extract, 5 g/L

technique(s)

microbiological culture: suitable

application(s)

agriculture
food and beverages
microbiology

storage temp.

room temp

suitability

nonselective for Escherichia coli
nonselective for coliforms

General description

Lennox LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides, amino acids, water-soluble vitamins, and carbohydrates in a low-salt formulation. The addition of agar provides a solid medium for microbial growth.

Application

Suitable for non-selective cultivation of E. coli strains for cloning, DNA plasmid production and production of recombinant proteins. Also suitable for selective cultivation when appropriate antibiotics are added, including those that require low-salt conditions, such as Zeocin®.

Features and Benefits

Lennox EZMix powder with agar provides:
  • A budget-friendly alternative to pre-poured plates
  • Granulated, dust-free format for safer handling and faster mixing
  • Convenient small package to eliminate weighing
  • Easy scale-up using larger package sizes
  • Standard formulation

Preparation Note

1. Suspend 35.6 g in 1 L of distilled water.
2. Heat to boiling while stirring to dissolve all ingredients completely.
3. Autoclave for 15 minutes at 121°C.
To prepare Lennox L Agar: Add 1 g glucose and proceed with preparation instructions as above.
To prepare the medium of Enquist and Sternberg: Aseptically add 10 ml of sterile 1 M magnesium sulfate after autoclaving.

Reconstitution

Add 35.6g powder in 1L water. Heat to boiling while stirring to dissolve powder. Autoclave for 15 minutes at 121C to sterilize. Allow to cool slightly before making additions, such as antibiotics (if desired). Pour into petri dishes and allow to solidify.

Legal Information

Zeocin is a registered trademark of Cayla Sarl

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sambrook, J., et al
Molecular Cloning: A Laboratory Manual, 2, 1-1 (1989)
Ausubel, F. M.
Current Protocols in Molecular Biology, 1, 3-3 (1994)
Alexis Kaushansky et al.
Nature protocols, 5(4), 773-790 (2010-04-03)
Protein microarrays provide an efficient way to identify and quantify protein-protein interactions in high throughput. One drawback of this technique is that proteins show a broad range of physicochemical properties and are often difficult to produce recombinantly. To circumvent these
Shuang Wang et al.
STAR protocols, 2(2), 100440-100440 (2021-04-27)
Studies to identify genes relevant to mammalian hepatocyte biology in vivo are largely carried out using germline genetic-engineering approaches, which can be costly and time-consuming. We describe hydrodynamic tail vein injection as an alternative approach to introduce genetic elements into
Raymond Liang et al.
Cell stem cell, 26(3), 359-376 (2020-02-29)
Quiescence is a fundamental property that maintains hematopoietic stem cell (HSC) potency throughout life. Quiescent HSCs are thought to rely on glycolysis for their energy, but the overall metabolic properties of HSCs remain elusive. Using combined approaches, including single-cell RNA

Questions

1–5 of 5 Questions  
  1. Will adding magnesium to the culture medium increase cell density?

    1 answer
    1. In microbial broth formulations that do not already contain magnesium, the addition of 10-20 mM MgCl2 or MgSO4 may increase cell densities.  You may need to also increase the shaking speed of the incubator.

      Helpful?

  2. Which bacterial culture medium is the best choice for my application?

    1 answer
    1. Each of the broths will likely grow E. coli very well, but there are still general guidelines for choosing a broth when you are working without a protocol. Generally:LB - Miller and LB - Lennox are used for E. coli growth and maintanence, DNA plasmid production and protein production.  The Lennox formulation has a lower salt content required for some salt-sensitive selection antibiotics.LB - Luria low salt is used for special applications where the E. coli growth or other constraints require the lowest possible salt content.Terrific Broth is used for higher yield protein production and high yield DNA plasmid production, because of the faster growth of the E. coli in this medium.SOB is used for protein production, DNA plasmid production and the generation of high-efficiency competent cells.SOC is used for initial growth of competent cells and the transformation procedure.

      Helpful?

  3. What are the differences among the Luria, Lennox and Miller LB formulations?

    1 answer
    1. LB, (originally termed lysogeny broth) was initially composed of tryptone, yeast extract, NaCl and glucose.  Soon after, the glucose was omitted (Miller's version), and later the NaCl content lowered by half (Lennox's version).  For some applications, even lower salt is required (Luria's low salt version).

      Helpful?

  4. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  5. What is the difference among the many LB - Lennox products?

    1 answer
    1. LB - Lennox is available in many types to suit your needs.  The different product formats include powder, EZMix™ powder, and tablet form.  Each of these forms are available with and without agar for easy LB-agar plate preparation.L3022 (LB Lennox powder)L2897 (LB Lennox powder with agar)L7275 (LB Lennox tablets)L7025 (LB Lennox tablets with agar)L7658 (LB Lennox EZMix(TM) powder)L7533 (LB Lennox EZMix(TM) powder with agar)L5542 (LB Lennox pre-poured agar plates)

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service