Skip to Content
Merck
All Photos(1)

Key Documents

J4829

Sigma-Aldrich

JFD00244

≥98% (HPLC), solid

Synonym(s):

1,4-Bis[(p-hydroxyphenethyl)amino]-anthraquinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H26N2O4
CAS Number:
Molecular Weight:
478.54
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Quality Level

Assay

≥98% (HPLC)

form

solid

color

violet

solubility

DMSO: ≥10 mg/mL
H2O: <2 mg/mL

storage temp.

room temp

SMILES string

Oc1ccc(CCNc2ccc(NCCc3ccc(O)cc3)c4C(=O)c5ccccc5C(=O)c24)cc1

InChI

1S/C30H26N2O4/c33-21-9-5-19(6-10-21)15-17-31-25-13-14-26(32-18-16-20-7-11-22(34)12-8-20)28-27(25)29(35)23-3-1-2-4-24(23)30(28)36/h1-14,31-34H,15-18H2

InChI key

UUJHFIBEJJLZBF-UHFFFAOYSA-N

Biochem/physiol Actions

JFD00244 is a SIRT (sirtuin, human silent information regulator) inhibitor.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anu J Tervo et al.
Journal of medicinal chemistry, 47(25), 6292-6298 (2004-11-30)
Type 2 human sirtuin (SIRT2) is a NAD(+)-dependent cytoplasmic protein that is colocalized with HDAC6 on microtubules. SIRT2 has been shown to deacetylate alpha-tubulin and to control mitotic exit in the cell cycle. To date, some small molecular inhibitors of
Anu J Tervo et al.
Journal of medicinal chemistry, 49(24), 7239-7241 (2006-11-28)
A successful virtual screening experiment of novel SIRT2 inhibitors is described. Four out of 11 experimentally tested compounds showed in vitro inhibitory activity toward SIRT2 in a micromolar level, resulting in an experimental hit ratio of 36%. Two of these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service