Skip to Content
Merck
All Photos(1)

Key Documents

I7908

Sigma-Aldrich

IL-2 human

≥98% (SDS-PAGE and HPLC), recombinant, expressed in Pichia pastoris, lyophiilized powder, suitable for cell culture

Synonym(s):

IL-2, Interleukin-2, T-cell growth factor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.75

product name

Interleukin-2 human, recombinant, expressed in Pichia pastoris, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in Pichia pastoris

Assay

≥98% (SDS-PAGE and HPLC)

form

lyophilized powder

mol wt

predicted mol wt ~15 kDa

packaging

pkg of 10KU

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... IL2(3558)

Looking for similar products? Visit Product Comparison Guide

Application

Interleukin-2 human has been used to assess changes in tumor necrosis factor receptor 1 (TNFR1). It has also been used in TNFR2 expression on Tregs.

Biochem/physiol Actions

Interleukin 2 (IL2) is a pro-inflammatory cytokine. It consists of three subunits, in which the β subunit is important for receptor mediated signaling. IL2 enhances the development of T regulatory cells and plays a crucial role in antitumor immunity.
Interleukin-2 (IL-2) is an immunomodulatory factor produced by certain subsets of T lymphocytes. This lympho­kine promotes long term growth of acti­vated T cells and related cell types. IL-2 plays a role in the activation and proliferation of NK cells, induces γ-inter­feron and B cell growth factor secretion, and modulates the expression of the IL-2 receptor.

Physical form

Lyophilized from a 0.2 μm filtered solution in 50 mM sodium phosphate buffer, pH 7.0, containing 0.1% human serum albumin per 0.1 mL of cytokine.

Analysis Note

The biological activity is tested in a cell proliferation assay using an IL-2 dependent murine cytotoxic T cell line, CTLL-2.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Racha Bahri et al.
Cancers, 14(12) (2022-06-25)
The Epstein-Barr virus (EBV) is associated with angioimmunoblastic T cell lymphoma (AITL), a peripheral T lymphoma of poor prognosis in at least 90% of cases. The role of EBV in this pathology is unknown. Using next-generation sequencing, we sequenced the
TNFR1 and TNFR2 expression and induction on human Treg cells from type 1 diabetic subjects
Heinrich M, et al.
Antibodies, 4(1), 34-47 (2015)
Meera V Singh et al.
Journal of leukocyte biology, 109(4), 807-820 (2020-07-15)
In addition to their role in hemostasis, platelets store numerous immunoregulatory molecules such as CD40L, TGFβ, β2-microglobulin, and IL-1β and release them upon activation. Previous studies indicate that activated platelets form transient complexes with monocytes, especially in HIV infected individuals
Nader Bayda et al.
Cancers, 13(4) (2021-02-10)
The Epstein-Barr virus (EBV) is associated with angioimmunoblastic T cell lymphoma (AITL) in more than 80% of cases. Few studies have focused on this association and it is not clear now what role the virus plays in this pathology. We
Fang Wang et al.
Journal of cellular and molecular medicine, 25(21), 10257-10267 (2021-10-22)
Oral lichen planus (OLP) is a T cell-mediated immunoinflammatory disease. Glycolysis plays an essential role in T-cell immune responses. Blocking glycolytic pathway in activated T cells represents a therapeutic strategy for restraint of immunologic process in autoimmune disorders. 2-Deoxy-D-glucose (2-DG)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service