Skip to Content
Merck
All Photos(1)

Key Documents

I117

Sigma-Aldrich

R(+)-IAA-94

≥98% (HPLC), solid

Synonym(s):

Indanyloxyacetic acid 94, R(+)-Methylindazone, R(+)-[(6,7-Dichloro-2-cyclopentyl-2,3-dihydro-2-methyl-1-oxo-1H-inden-5-yl)-oxy]acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18Cl2O4
CAS Number:
Molecular Weight:
357.23
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

optical activity

[α]22/D +36.4°, c = 0.86 in methanol(lit.)

color

white

solubility

H2O: 0.2 mg/mL
0.1 M NaOH: 2.8 mg/mL
DMSO: >20 mg/mL
ethanol: 20 mg/mL
dilute aqueous acid: insoluble

storage temp.

2-8°C

SMILES string

C[C@@]1(Cc2cc(OCC(O)=O)c(Cl)c(Cl)c2C1=O)C3CCCC3

InChI

1S/C17H18Cl2O4/c1-17(10-4-2-3-5-10)7-9-6-11(23-8-12(20)21)14(18)15(19)13(9)16(17)22/h6,10H,2-5,7-8H2,1H3,(H,20,21)/t17-/m1/s1

InChI key

RNOJGTHBMJBOSP-QGZVFWFLSA-N

Application

R(+)-IAA-94 has been used as a chloride ion channel inhibitor to study its effects on grepafloxacin uptake by human mononuclear (THP-1) cells . It has also been used as a chloride ion channel inhibitor to study its effects on moxifloxacin uptake by human mononuclear (THP-1) cells.

Features and Benefits

This compound is featured on the Chloride Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D W Landry et al.
The Journal of general physiology, 90(6), 779-798 (1987-12-01)
Chloride channels are present in the majority of epithelial cells, where they mediate absorption or secretion of NaCl. Although the absorptive and secretory channels are well characterized in terms of their electrophysiological behavior, there is a lack of pharmacological ligands
Human THP-1 monocyte uptake and cellular disposition of 14C-grepafloxacin
Hall I H, et al.
Journal of Infection and Chemotherapy, 10(1), 11-18 (2004)
D M Briggs Boedtkjer et al.
Pflugers Archiv : European journal of physiology, 457(2), 389-404 (2008-06-10)
The possibility that Ca(2+)-activated Cl(-) conductances (CaCCs) contribute to oscillations in vascular tone (vasomotion) is tested in isolated mesenteric small arteries from rats where cGMP independent (I (Cl(Ca))) and cGMP-dependent (I (Cl(Ca,cGMP))) chloride conductances are important. The effect of anion
Michelle Batthish et al.
Cardiovascular research, 55(3), 660-671 (2002-08-06)
We have recently proposed that chloride (Cl(-)) channels contribute to ischemic preconditioning (IPC) in the myocardium. To further evaluate this hypothesis, we investigated the role of Cl(-) channels in pharmacological preconditioning. Isolated rabbit cardiomyocytes and isolated buffer-perfused rabbit hearts were
Influence of T-type Ca2+ (mibefradil) and Cl- (indanyloxyacetic acid 94) channel antagonists on ?1-adrenoceptor mediated contractions in rat aorta.
Duggan AJ and Tabrizchi R
Canadian Journal of Physiology and Pharmacology, 78(9), 714-720 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service