Skip to Content
Merck
All Photos(4)

Key Documents

HPA053254

Sigma-Aldrich

Anti-LVRN antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-APQ, Anti-AQPEP, Anti-FLJ90650, Anti-TAQPEP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:2500- 1:5000

immunogen sequence

IPYPIKDVVLCYGIALGSDKEWDILLNTYTNTTNKEEKIQLAYAMSCSKDPWILNRYMEYAISTSPFTSNETNIIEVVASSEVGRYVA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LVRN(206338)

General description

Laeverin (LVRN), also called aminopeptidase Q, is a 120 kDa protein, which belongs to M1 family of aminopeptidases. It is a type II integral membrane protein with HEXXHX18E gluzincin motif. It is specifically expressed in extravillous trophoblast of placenta. The LVRN gene is localized on human chromosome 5q23.1.

Immunogen

laeverin

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Laeverin (LVRN) is crucial for human placentation and maintenance of pregnancy in humans. The catalytic pocket residue histidine at position 379, is crucial for its functionality. The glutamine residue at the 238 position, is essential for electrostatic interaction with substrate. It plays a key role in extravillous trophoblast migration. It is highly expressed in preeclamptic placenta and may be involved in the pathogenesis of preeclampsia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84454

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laeverin/aminopeptidase Q induces trophoblast invasion during human early placentation
Horie A, et al.
Human Reproduction, 27(5), 1267-1276 (2012)
Histidine-379 of human laeverin/aminopeptidase Q, a non-conserved residue within the exopeptidase motif, defines its distinctive enzymatic properties
Maruyama M, et al.
The Journal of Biological Chemistry, 284(50), 34692?34702-34692?34702 (2009)
Common genes underlying asthma and COPD? Genome-wide analysis on the Dutch hypothesis
Smolonska J, et al.
The European Respiratory Journal, 211(6), erj00019-erj02014 (2014)
Involvement of glutamine-238 in the substrate specificity of human laeverin/aminopeptidase Q
Goto Y, et al.
Biological & Pharmaceutical Bulletin, 34(1), 24-27 (2011)
Placental expression of aminopeptidase-Q (laeverin) and its role in the pathophysiology of preeclampsia
Nystad M, et al.
American Journal of Obstetrics and Gynecology, 211(6), 686-e1-686-e1 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service