Skip to Content
Merck
All Photos(3)

Key Documents

ABN302

Sigma-Aldrich

Anti-Complex I-75kD Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

NADH-ubiquinone oxidoreductase 75 kDa subunit, mitochondrial, Complex I-75kD, CI-75kD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat, human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), chimpanzee (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NDUFS1(4719)

General description

Complex I-75kD (CI-75kD) is also called NADH-ubiquinone oxidoreductase 75 kDa subunit, mitochondrial (NDUFS1). Complex I-75kD is a core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), which transfers electrons from NADH to the respiratory chain, and is thought to be part of the minimal assembly required for catalysis. Complex I-75kD defects are thought to be a cause of Mitochondrial complex I deficiency (MT-C1D).

Immunogen

KLH-conjugated linear peptide corresponding to 15 amino acids from the internal region of human Complex I-75kD.

Application

Anti-Complex I-75kD, Cat. No. ABN302, is a rabbit polyclonal antibody that detects Complex I-75kD and is tested for use in Western Blotting and Immunohistochemistry.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling
Tested Applications

Western Blotting Analysis: A 1:5,000 dilution from a representative lot detected Complex I-75kD in rat brain, human brain, and human heart tissue lysates.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution of this antibody detected Complex I-75kD in human kidney and human heart tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Quality

Evaluated by Western Blotting in mouse brain tissue lysate.

Western Blotting Analysis: A 1:5,000 dilution of this antibody detected Complex I-75kD in mouse brain tissue lysates.

Target description

~75 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leena P Bharath et al.
Cell metabolism, 32(1), 44-55 (2020-05-14)
Age is a non-modifiable risk factor for the inflammation that underlies age-associated diseases; thus, anti-inflammaging drugs hold promise for increasing health span. Cytokine profiling and bioinformatic analyses showed that Th17 cytokine production differentiates CD4+ T cells from lean, normoglycemic older and
Lauren G Buss et al.
Scientific reports, 14(1), 845-845 (2024-01-09)
Salivary glands are indirectly damaged during radiotherapy for head and neck cancer, resulting in acute and chronic hyposalivation. Current treatments for radiation-induced hyposalivation do not permanently restore function to the gland; therefore, more mechanistic understanding of the damage response is
Vedavathi Madhu et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 35(8), 1504-1524 (2020-04-07)
Nucleus pulposus (NP) cells reside in an avascular and hypoxic microenvironment of the intervertebral disc and are predominantly glycolytic due to robust HIF-1 activity. It is generally thought that NP cells contain few functional mitochondria compared with cells that rely
Caroline A Kopsidas et al.
iScience, 27(7), 110199-110199 (2024-07-11)
Neurons in the neocortex are generated during embryonic development. While the adult ventricular-subventricular zone (V-SVZ) contains cells with neural stem/progenitors' characteristics, it remains unclear whether it has the capacity of producing neocortical neurons. Here, we show that generating neurons with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service