Skip to Content
Merck
All Photos(8)

Key Documents

HPA017319

Sigma-Aldrich

Anti-IFNLR1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CRF2-12, Anti-Cytokine receptor class-II member 12, Anti-Cytokine receptor family 2 member 12, Anti-IFN-lambda R1, Anti-IL-28R-alpha, Anti-IL-28RA, Anti-IL28RA, Anti-Interferon lambda receptor 1, Anti-Interleukin-28 receptor alpha chain precursor, Anti-Likely interleukin or cytokine receptor 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

FEVEPAPPVLVLTQTEEILSANATYQLPPCMPPLDLKYEVAFWKEGAGNKTLFPVTPHGQPVQITLQPAASEHHCLSARTIYTFSVPKYSKFSKPTCFLLEVPEAN

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL28RA(163702)

General description

IFNLR1 (Interferon λ receptor 1) is an orphan class II receptor chain belonging to the heterodimeric class II cytokine receptor. It is widely expressed in human cell lines and tissues.

Immunogen

Interleukin-28 receptor alpha chain precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-IL28RA antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

IFNLR1 (Interferon λ receptor 1) functions as a component of IFN-λ protein. It forms a ligand-receptor system upon binding to interferon-λ1. It actively participates in the JAK-STAT (Janus kinases-signal transducers and activators of transcription) pathway. It has been suggested that IFNLR1 polymorphism can be an effective tool for identifying early treatment failure of early hepatitis C virus (HCV) 2 treatment.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72355

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Jiménez-Sousa et al.
Journal of viral hepatitis, 20(5), 358-366 (2013-04-10)
Due to the poor rate of response to hepatitis C virus (HCV) with pegylated interferon and ribavirin treatment in HCV/HIV coinfected patients, key factors for predicting failure would be useful. We performed a retrospective study on 291 patients on HCV
Paul Sheppard et al.
Nature immunology, 4(1), 63-68 (2002-12-07)
Cytokines play a critical role in modulating the innate and adaptive immune systems. Here, we have identified from the human genomic sequence a family of three cytokines, designated interleukin 28A (IL-28A), IL-28B and IL-29, that are distantly related to type
Rama K Mallampalli et al.
Frontiers in immunology, 12, 735576-735576 (2021-12-14)
Interferon lambda (IFNλ) signaling is a promising therapeutic target against viral infection in murine models, yet little is known about its molecular regulation and its cognate receptor, interferon lambda receptor 1 (IFNLR1) in human lung. We hypothesized that the IFNλ
Sabine Zahn et al.
The Journal of investigative dermatology, 131(1), 133-140 (2010-08-20)
Type I IFNs (IFNα/β) have been shown to have a central role in the pathophysiology of lupus erythematosus (LE). The recently discovered type III IFNs (IFNλ1/IL29, IFNλ2/IL28a, IFNλ3/IL28b) share several functional similarities with type I IFNs, particularly in antiviral immunity.
Oliver Bauhofer et al.
Gastroenterology, 143(2), 429-438 (2012-04-24)
Hepatitis C virus (HCV) is a common cause of chronic liver disease. Many patients do not clear the viral infection; little is known about the mechanisms of HCV persistence or the frequent failure of interferon (IFN) to eliminate it. Better

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service