Skip to Content
Merck
All Photos(8)

Key Documents

HPA015768

Sigma-Aldrich

Anti-S100B Antibody

enhanced validation

rabbit polyclonal

Synonym(s):

Anti-Protein S100-B, Anti-S-100 protein beta chain, Anti-S-100 protein subunit beta, Anti-S100 calcium-binding protein B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

Product Name

Anti-S100B antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

LEKAMVALIDVFHQYSGREGDKHKLKKSELKELINNELSHFLEEIKEQEVVDKVMETLDNDGDGECDFQEFMAFVAMVTTACHEFFEHE

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... S100B(6285)

General description

S100B is a Ca2+ binding protein that is secreted by the astrocytes by an autocrine pathway to activate the release of nitric oxide. S100B may regulate the functions of microglial cells . It may also be associated with the dynamics of intermediary filaments and microtubules in glial cells . Anti-S100B antibodies are specific for S100B in humans. The gene encoding S100B is localized on human chromosome 21.

Immunogen

Protein S100-B recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunocytochemistry (1 paper)
Immunofluorescence (1 paper)
Immunohistochemistry (1 paper)

Biochem/physiol Actions

S-100 β subunit (S100B) exerts paracrine and autocrine effects on neurons and glia. At nanomolar concentrations, the encoded protein promotes neurite outgrowth and increases survival of neurons during development. S100B serves as a marker of melanocyte cytotoxicity. In addition, it also acts as a serum marker in endocrine resistant breast cancer and nonsegmental vitiligo. Decreased expression of S100B has been observed in chronic liver disease patients.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73328

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lifan Zhu et al.
Molecular medicine reports, 18(6), 4855-4864 (2018-10-04)
The present study aimed to investigate the role of S100B in the inflammation process during osteoarthritis (OA). OA cartilage samples were collected for S100B expression analysis. S100B expression levels were significantly increased in patients with OA compared with the Controls
Rapid detection of trisomy 21
by homologous gene quantitative PCR (HGQ-PCR)
Lee H, et al.
Human Genetics, 99, 364-367 (1997)
Association of S100B with intermediate filaments and microtubules in glial cells
Sorci, G., et al.
Biochimica et Biophysica Acta - Molecular Cell Research, 1448(2), 277-289 (1998)
Siobhan S Pattwell et al.
Nature communications, 7, 11475-11475 (2016-05-25)
Fear can be highly adaptive in promoting survival, yet it can also be detrimental when it persists long after a threat has passed. Flexibility of the fear response may be most advantageous during adolescence when animals are prone to explore
Elma Aflaki et al.
Neurobiology of disease, 134, 104647-104647 (2019-11-02)
While astrocytes, the most abundant cells found in the brain, have many diverse functions, their role in the lysosomal storage disorder Gaucher disease (GD) has not been explored. GD, resulting from the inherited deficiency of the enzyme glucocerebrosidase and subsequent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service