Skip to Content
Merck
All Photos(1)

Key Documents

MAB356

Sigma-Aldrich

Anti-Substance P Antibody, pain, clone NC1

culture supernatant, clone NC1, Chemicon®

Synonym(s):

Anti-Hs.2563, Anti-NK2, Anti-NKNA, Anti-NPK, Anti-TAC2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

NC1, monoclonal

species reactivity

human

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TAC1(6863)

Specificity

Reacts with COOH-terminal end of Substance P. No cross-reactivity to Leu- or Met- enkephalin, somatostatin or beta-endorphin. Eledoisin 5%.

Immunogen

Substance P conjugated to BSA.

Application

1:100 - 1:200 by immunohistochemistry on human brain.

Optimal working dilutions must be determined by end user.
Detect Substance P using this Anti-Substance P Antibody, pain, clone NC1 validated for use in IH.

Physical form

Lyophilized Supernatant, preservative free.

Storage and Stability

Lyophilized antibody can be kept at 2-8°C for up to 3 months and should be kept at -20°C for long term storage. Reconstitute with 100μL sterile DI water. Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liang Han et al.
Nature neuroscience, 21(3), 324-328 (2018-02-07)
Asthma, accompanied by lung inflammation, bronchoconstriction and airway hyper-responsiveness, is a significant public health burden. Here we report that Mas-related G protein-coupled receptors (Mrgprs) are expressed in a subset of vagal sensory neurons innervating the airway and mediates cholinergic bronchoconstriction
High-fat diet: bacteria interactions promote intestinal inflammation which precedes and correlates with obesity and insulin resistance in mouse.
Ding, S; Chi, MM; Scull, BP; Rigby, R; Schwerbrock, NM; Magness, S; Jobin, C; Lund, PK
Testing null
Andrew P Ray et al.
Pharmacology, biochemistry, and behavior, 94(1), 211-218 (2009-08-25)
Research on the mechanisms of emesis has implicated multiple neurotransmitters via both central (dorsal vagal complex) and peripheral (enteric neurons and enterochromaffin cells) anatomical substrates. Taking advantage of advances in receptor-specific agonists, and utilizing Fos expression as a functional activity
Hypoalgesia in mice with a targeted deletion of the tachykinin 1 gene.
Zimmer, A; Zimmer, AM; Baffi, J; Usdin, T; Reynolds, K; Konig, M; Palkovits, M; Mezey, E
Proceedings of the National Academy of Sciences of the USA null
Tibor Z Veres et al.
American journal of respiratory cell and molecular biology, 37(5), 553-561 (2007-06-30)
Neuroimmune interactions play a critical role in the pathogenesis of asthma. Symptoms like wheezing and cough have been attributed to neural dysregulation, whereas sensitization and the induction of allergic inflammation have been linked with the activity of dendritic cells. Neuropeptides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service