Skip to Content
Merck
All Photos(2)

Documents

HPA013132

Sigma-Aldrich

Anti-OGN antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Mimecan precursor, Anti-OIF, Anti-Osteoglycin, Anti-Osteoinductive factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

SLLEELSLAENQLLKLPVLPPKLTLFNAKYNKIKSRGIKANAFKKLNNLTFLYLDHNALESVPLNLPESLRVIHLQFNNIASITDDTFCKANDTSYIRDRIEEIRLEGNPIVLGKHPNSFI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OGN(4969)

General description

Osteoglycin (OGN) is a 22-28kDa glycoprotein which is present in the bone matrix. It is heavily glycosylated. OGN belongs to the small leucine-rich proteoglycans (SLRPs) family. The gene encoding it is localized on chromosome 9q22.

Immunogen

Mimecan precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Osteoglycin (OGN) has been shown to induce ectopic bone formation. It can stimulate the proliferation of osteoblastic cells and enhances their alkaline phosphate activity. OGN also inhibits total multinucleated cell (MNC) formation. It binds to transforming growth factor β (TGF-β), which has a role in vascular smooth muscle disease. OGN may be a marker for diagnosing pituitary tumors.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71483

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin M Cheng et al.
Arteriosclerosis, thrombosis, and vascular biology, 34(5), 1078-1084 (2014-03-22)
Previous proteomics experiments have demonstrated that several proteins are differentially expressed in vulnerable human carotid plaques compared with stable plaques. This study aims to investigate the prognostic value of 13 such circulating biomarkers in patients with coronary artery disease. Between
Yutaka Yabe et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 34(7), 1241-1247 (2015-12-19)
Elastic fibers and proteoglycans are major components of the extracellular matrix and their changes have been reported in some pathological conditions. Further, recent studies have indicated that some glycosaminoglycans and proteoglycans inhibit elastic fiber assembly. The purpose of this study
A Kukita et al.
Proceedings of the National Academy of Sciences of the United States of America, 87(8), 3023-3026 (1990-04-01)
Osteoinductive factor (OIF) is a glycoprotein in bone that induces ectopic bone formation. Implantation of OIF plus transforming growth factor beta (TGF-beta) type 1 or 2 into subcutaneous tissues of rats induces formation of bone at the implantation site. Since
Bonan Chen et al.
Clinical and translational medicine, 13(12), e1516-e1516 (2023-12-27)
Cancer-associated fibroblasts (CAFs), integral to the tumour microenvironment, are pivotal in cancer progression, exhibiting either pro-tumourigenic or anti-tumourigenic functions. Their inherent phenotypic and functional diversity allows for the subdivision of CAFs into various subpopulations. While several classification systems have been
Assignment of mimecan gene (OGN) to human chromosome band 9q22 by in situ hybridization.
E S Tasheva et al.
Cytogenetics and cell genetics, 88(3-4), 326-327 (2000-06-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service