Skip to Content
Merck
All Photos(6)

Key Documents

HPA012947

Sigma-Aldrich

Anti-STMN3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-SCG10-like protein, Anti-Stathmin-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:500-1:1000
western blot: 0.04-0.4 μg/mL

immunogen sequence

TQEAQVLKQLAERREHEREVLHKALEENNNFSRQAEEKLNYKMELSKEIREAHLAALRERLREKELHAAEVRRNKEQREEMSG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STMN3(50861)

General description

Stathmin-3 (STMN3), a 180 amino acid protein, belongs to the stathmin family of phospho-proteins. It is localized in the Golgi complex and is seen in the vesicles along axons and dendrites.

Immunogen

Stathmin-3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Stathmin-3 (STMN3) is mainly involved in regulating dendritic formation and axonal branching. It interacts with Ras protein-specific guanine nucleotide-releasing factor 1 (RasGRF1) and suppresses RasGRF1-mediated activation of Rac (GTPase)/p38 mitogen-activated protein kinases (MAPK) pathway. The Dbl-homology (DH) domain of RasGRF1 is responsible for its exchange activity on Rac. STMN3 association with RasGRF1 masks this DH domain. It is largely known as a microtubule-destabilizing protein which prevents the polymerization of microtubules by sequestering soluble tubulin heterodimers. High levels of STMN3 are observed in metastatic ovarian cancer tissue.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71127

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuemei Xie et al.
Molecular cancer therapeutics, 12(6), 1099-1111 (2013-04-02)
Paclitaxel is a standard chemotherapeutic agent for ovarian cancer. PEA-15 (phosphoprotein enriched in astrocytes-15 kDa) regulates cell proliferation, autophagy, apoptosis, and glucose metabolism and also mediates AKT-dependent chemoresistance in breast cancer. The functions of PEA-15 are tightly regulated by its
Jennifer Walter-Yohrling et al.
Cancer research, 63(24), 8939-8947 (2003-12-26)
To systematically identify genes related to invasion a three-dimensional multicellular matrix invasion assay was used to classify human tumor cell lines as stromal invasion positive or stromal invasion negative. Cells from two of the primary cell types of the stromal
Simona Baldassa et al.
The Journal of biological chemistry, 282(4), 2333-2345 (2006-12-01)
RasGRF1 is a neuron-specific guanine nucleotide exchange factor for the small GTPases Ras and Rac. It is implicated in the regulation of memory formation and in the development of tolerance to drug abuse, although the mechanisms have been elucidated only
Matthew A Helsby et al.
BMC cell biology, 15, 6-6 (2014-02-18)
Research antibodies are used by thousands of scientists working in diverse disciplines, but it is common to hear concerns about antibody quality. This means that researchers need to carefully choose the antibodies they use to avoid wasting time and money.
Aurore D Levy et al.
Molecular biology of the cell, 22(11), 1930-1942 (2011-04-08)
Protein palmitoylation is a reversible lipid modification that plays critical roles in protein sorting and targeting to specific cellular compartments. The neuronal microtubule-regulatory phosphoproteins of the stathmin family (SCG10/stathmin 2, SCLIP/stathmin 3, and RB3/stathmin 4) are peripheral proteins that fulfill

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service