Skip to Content
Merck
All Photos(6)

Documents

HPA014855

Sigma-Aldrich

Anti-DDX47 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DEAD box protein 47, Anti-Probable ATP-dependent RNA helicase DDX47

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

IHRVGRTARAGRSGKAITFVTQYDVELFQRIEHLIGKKLPGFPTQDDEVMMLTERVAEAQRFARMELREHGEKKKRSREDAGDNDDTEGAIGVRNKVAGGKMKKRKGR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DDX47(51202)

General description

DDX47 (DEAD (Asp-Glu-Ala-Asp) box polypeptide 47) is a member of the DEAD box protein family, and shares homology with the members at its N-terminal. It has the eight motifs characteristic of DEAD box member proteins, such as Walker A and Walker B motifs. It resides in the nucleolus, but shuttles between cytoplasm and nucleus. During cell division, it is found almost entirely in the cytoplasm. It has a molecular weight of 66kDa. It has a wide range of tissue expression including retina, liver, testis, placenta and T-cells.

Immunogen

Probable ATP-dependent RNA helicase DDX47 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The exact function of DDX47 (DEAD (Asp-Glu-Ala-Asp) box polypeptide 47) is not well understood, but it is thought that this protein plays a role in the biosynthesis of ribosomes. This protein interacts with the C-terminal of the E1E4 protein of human papillomavirus type 16 (HPV16), and might be involved in the replication of HPV16. This protein is also a binding partner of GABA(A) receptor-associated protein (GABARAP), which acts a chaperone for GABA (γ-aminobutyric aid)(A) receptor in the neurons of cortex. Together, GABARAP and DDX47 might be involved in apoptosis. DDX47 is involved in the processing of pre-rRNA, and is recruited to the pre-rRNA processing region by NOP132 (nucleolar protein).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73083

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeong Heon Lee et al.
Biotechnology letters, 27(9), 623-628 (2005-06-25)
GABA(A) receptor-associated protein (GABARAP) is a 14-kDa cytoplasmic protein initially identified as a molecular chaperone for GABA(A) receptor in cortical neurons. However, evidence indicates that the function of GABARAP is much broader than a specific role in neuronal cells. As
J Doorbar et al.
Journal of virology, 74(21), 10081-10095 (2000-10-12)
Human papillomavirus type 16 (HPV16) infects cervical epithelium and is associated with the majority of cervical cancers. The E1E4 protein of HPV16 but not those of HPV1 or HPV6 was found to associate with a novel member of the DEAD
Takeshi Sekiguchi et al.
Nucleic acids research, 34(16), 4593-4608 (2006-09-12)
Previously, we described a novel nucleolar protein, NOP132, which interacts with the small GTP binding protein RRAG A. To elucidate the function of NOP132 in the nucleolus, we identified proteins that interact with NOP132 using mass spectrometric methods. NOP132 associated
Esther Marchena-Cruz et al.
Cell reports, 42(3), 112148-112148 (2023-02-25)
Unscheduled R loops can be a source of genome instability, a hallmark of cancer cells. Although targeted proteomic approaches and cellular analysis of specific mutants have uncovered factors potentially involved in R-loop homeostasis, we report a more open screening of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service