Skip to Content
Merck
All Photos(7)

Key Documents

HPA003259

Sigma-Aldrich

Anti-MITF antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

MI, Microphthalmia-associated transcription factor, WS2, WS2A, bHLHe32

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

HLLLRIQELEMQARAHGLSLIPSTGLCSPDLVNRIIKQEPVLENCSQDLLQHHADLTCTTTLDLTDGTITFNNNLGTGTEANQAYSVPTKMGSKLEDILMDDTLSPVGVTDPLLSSVSPGASKTSSRRSSMSMEETEHT

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MITF(4286)

General description

MITF (microphthalmia-associated transcription factor) is a basic helix-loop-helix, leucine-zipper transcription factor. It is located on human chromosome 3p13.
Microphthalmia-associated transcription factor (MITF) is a bHLH-ZIP transcription factor that recognizes E-box (CAYRTG) and M-box (TCAYRTG or CAYRTGA) sequences in the promoter regions of target genes. Microphthalmia-associated transcription factor (MITF) is a master regulator in melanocyte proliferation, development, survival and melanoma formation and an essential regulator of osteoclastogenesis.
Rabbit polyclonal anti-MITF antibody recognizes human microphthalmia-associated transcription factor.

Immunogen

Microphthalmia-associated transcription factor recombinant protein epitope signature tag (PrEST)

Application

Anti-MITF antibody has been used in immunohistochemistry and chromatin immunoprecipitation.
Rabbit polyclonal anti-MITF antibody is used to tag microphthalmia-associated transcription factor for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of microphthalmia-associated transcription factor in melanocyte proliferation, development, and survival and the regulation of regulator of osteoclastogenesis.

Biochem/physiol Actions

MITF (microphthalmia-associated transcription factor) participates in the growth and differentiation of melanocytes. Mutations in MITF results in waardenburg syndrome type IIa.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84788

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefanie Riesenberg et al.
Nature communications, 6, 8755-8755 (2015-11-05)
Inflammation promotes phenotypic plasticity in melanoma, a source of non-genetic heterogeneity, but the molecular framework is poorly understood. Here we use functional genomic approaches and identify a reciprocal antagonism between the melanocyte lineage transcription factor MITF and c-Jun, which interconnects
The MITF, p. E318K Variant, as a Risk Factor for Pheochromocytoma and Paraganglioma.
Castro-Vega L J, et al.
The Journal of clinical endocrinology and metabolism, 101(12), 4764-4768 (2016)
Human melanocytes mitigate keratinocyte-dependent contraction in an in vitro collagen contraction assay
Rakar J, et al.
Burns : Journal of the International Society For Burn Injuries, 41(5), 1035-1042 (2015)
Melanocyte development in the mouse tail epidermis requires the Adamts9 metalloproteinase
Tharmarajah G, et al.
Pigment Cell & Melanoma Research (2018)
Grace Tharmarajah et al.
Pigment cell & melanoma research, 31(6), 693-707 (2018-05-22)
The mouse tail has an important role in the study of melanogenesis, because mouse tail skin can be used to model human skin pigmentation. To better understand the development of melanocytes in the mouse tail, we cloned two dominant ENU-generated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service