Skip to Content
Merck
All Photos(1)

Key Documents

284M-9

Sigma-Aldrich

Microphthalmia Transcription Factor (MiTF) (C5/D5) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

C5/D5, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (284M-94)
vial of 0.5 mL concentrate (284M-95)
bottle of 1.0 mL predilute (284M-97)
vial of 1.0 mL concentrate (284M-96)
bottle of 7.0 mL predilute (284M-98)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG1

control

melanoma

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

human ... MITF(4286)

General description

Mi is a transcription factor implicated in pigmentation, bone development and in mast cells. Various forms of Mi exist ranging from 50-70 kD in size. This antibody targets the 52-56 kD range. This antibody has been useful in identifying malignant melanoma, and distinguishing mast cell lesions from lesions of myeloid derivation. A relatively rare class of tumors known as PEComas (tumors showing perivascular epithelioid cell differentiation) express MiTF in a high percentage of cases (~90%).

Quality


IVD

IVD

IVD

RUO

Linkage

Microphthalmia Transcription Factor (MiTF) Positive Control Slides, Product No. 284S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bernadette Liegl et al.
The American journal of surgical pathology, 32(4), 608-614 (2008-02-19)
PEComas arising in somatic soft tissue or skin are rare. To further characterize the clinicopathologic spectrum, we herein report a series of 10 primary cutaneous PEComas. Eight patients were female and 2 patients were male. The age at presentation ranged
Steven J Ohsie et al.
Journal of cutaneous pathology, 35(5), 433-444 (2008-04-11)
Melanoma has a wide spectrum of histologic features which mimic epithelial, hematologic, mesenchymal, and neural tumors. Immunohistochemistry has been the primary tool to distinguish melanomas from these other tumors; it has also been studied for use as an adjunct to
Ilan Weinreb et al.
Virchows Archiv : an international journal of pathology, 450(4), 463-470 (2007-03-23)
Four cases of malignant PEComas were stained with smooth muscle actin, muscle specific actin, desmin, human melanoma black-45, melan-A, microphthalmia transcription factor, S100 and cyclin D1. One case was studied by electron microscopy (EM). Tumour locations were the thigh, elbow
Jason L Hornick et al.
The American journal of surgical pathology, 32(4), 493-501 (2008-01-29)
PEComas (tumors showing perivascular epithelioid cell differentiation) are a family of mesenchymal neoplasms that include angiomyolipoma, clear cell "sugar" tumor of the lung, lymphangiomyomatosis, and a group of uncommon lesions that arise in soft tissue, visceral organs, and skin. We
Alberto Righi et al.
International journal of surgical pathology, 16(1), 16-20 (2008-01-22)
We report 2 cases of PEComa, one occurring in the colon of an 11-year-old boy and the other in the bone (fibula) of a 92-year-old woman. Both tumors consisted of nests of large epithelioid cells surrounded by a fibrovascular stroma.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service