Skip to Content
Merck
All Photos(2)

Key Documents

HPA001220

Sigma-Aldrich

Anti-GAL3ST1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-3′-Phosphoadenosine-5′-phosphosulfate:GalCer sulfotransferase antibody produced in rabbit, Anti-3′-Phosphoadenylylsulfate:galactosylceramide 3′-sulfotransferase antibody produced in rabbit, Anti-Cerebroside sulfotransferase antibody produced in rabbit, Anti-GalCer sulfotransferase antibody produced in rabbit, Anti-Galactosylceramide sulfotransferase antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

LLNILFRFGQKHRLKFAFPNGRNDFDYPTFFARSLVQDYRPGACFNIICNHMRFHYDEVRGLVPTNAIFITVLRDPARLFESSFHYFGPVVPLTWKLSAGDKLTEFLQDPDRYYDPNGFNAHYLRNLLFFDLGYDNSLDPSSPQVQEHIL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GAL3ST1(9514)

Immunogen

Galactosylceramide sulfotransferase recombinant protein epitope signature tag (PrEST)

Application

Anti-GAL3ST1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

GAL3ST1 (Galactosylceramide sulfotransferase) gene encodes a protein that catalyzes the sulfonation of membrane glycolipids. It is also known as cerebroside sulfotransferase, CST. It catalyzes galactosylceramide sulfate (sulfatide) synthesis in the Golgi apparatus. Sulfatide is a major lipid component of the myelin sheath and of seminolipid present in spermatocytes. Its activity is increased in renal cancer cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73404

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Tsuda et al.
European journal of biochemistry, 267(9), 2672-2679 (2000-04-28)
The galactosylceramide sulfotransferase (cerebroside sulfotransferase, CST) (EC 2.8.2.11) gene is highly expressed in human renal cancer cells. To elucidate the regulatory mechanism of its gene expression, we have determined the genomic organization of the human CST gene. The gene comprises
Matthias Eckhardt et al.
The Biochemical journal, 368(Pt 1), 317-324 (2002-08-15)
3- O -Sulphogalactosylceramide (sulphatide) is a major lipid component of myelin membranes, and is required for proper myelin formation. Sulphatide is synthesized in the Golgi apparatus by galactosylceramide sulphotransferase (CST; EC 2.8.2.11). Murine and human CSTs contain two putative N-glycosylation
Michael A Kiebish et al.
Journal of lipid research, 53(2), 273-281 (2011-11-25)
Peroxisome proliferator-activated receptor gamma coactivator-1α (PGC-1α), a key regulator of energy metabolism and lipid homeostasis in multiple highly oxidative tissues, has been implicated in the metabolic derangements of diabetes and obesity. However, relatively less is known regarding its role in
K Honke et al.
Journal of biochemistry, 119(3), 421-427 (1996-03-01)
We have purified 3'-phosphoadenosine-5'-phosphosulfate:GalCer sulfotransferase [EC 2.8.2.11] from a human renal cancer cell line SMKT-R3 through a combination of affinity chromatographies using galactosylsphingosine, 3',5'-bisphosphoadenosine and heparin as ligands. The purified sulfotransferase showed a specific activity of 1.2 mumol/min/mg, which is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service