Skip to Content
Merck
All Photos(3)

Key Documents

H6883

Sigma-Aldrich

3-(N,N-Dimethylpalmitylammonio)propanesulfonate

≥98% (TLC)

Synonym(s):

3-(N,N-Dimethylhexadecylammonio)propanesulfonate, 3-(Hexadecyldimethylammonio)propanesulfonate, 3-(Palmityldimethylammonio)propanesulfonate, Palmityl sulfobetaine, SB3-16

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15N+(CH3)2CH2CH2CH2SO3-
CAS Number:
Molecular Weight:
391.65
Beilstein:
4149854
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

description

zwitterionic

Assay

≥98% (TLC)

mol wt

micellar avg mol wt 60,700

aggregation number

155

technique(s)

electrophoresis: suitable
mass spectrometry (MS): suitable
thin layer chromatography (TLC): suitable

conductivity

≤10 μmho per cm at 24 °C (0.05 M aqueous solution)

CMC

0.01-0.06 mM (20-25°C)

application(s)

cell analysis

SMILES string

CCCCCCCCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O

InChI

1S/C21H45NO3S/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19-22(2,3)20-18-21-26(23,24)25/h4-21H2,1-3H3

InChI key

TUBRCQBRKJXJEA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-(N,N-Dimethylpalmitylammonio)propanesulfonate is a zwitterionic detergent. [1]

Application

3-(N,N-Dimethylpalmitylammonio)propanesulfonate has been used in a study to assess the characterization of rat dihydroceramide desaturase. [2] It has also been used in a study to investigate the inhibition of mitochondrial carnitine–acylcarnitine translocase by sulfobetaines. [3] It is also used for extraction of proteins for analysis by chromatography, mass spectroscopy, and electrophoretic methods.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R J Barry et al.
The Journal of biological chemistry, 264(16), 9231-9238 (1989-06-05)
Studies reported previously from this laboratory have demonstrated that membranes from the pigment epithelium of the vertebrate eye can transform free all-trans-retinol to 11-cis-retinol as well as 11-cis- and all trans-retinyl esters (Bernstein, P. S., Law, W. C., and Rando
C Causeret et al.
Lipids, 35(10), 1117-1125 (2000-12-05)
The introduction of the double bond in the sphingoid backbone of sphingolipids occurs at the level of dihydroceramide via an NADPH-dependent desaturase, as discovered in permeabilized rat hepatocytes. In the rat, the enzyme activity, which has now been further characterized
H Erdjument-Bromage et al.
Journal of chromatography. A, 826(2), 167-181 (1999-01-01)
Mass spectrometry occupies a central position in most current protein identification schemes. So-called 'mass fingerprinting' techniques rely on composite mass patterns of proteolytic fragments, or dissociation products thereof, to query databases. Keys to successful analysis of ever smaller amounts are
R Parvin et al.
Canadian journal of biochemistry, 58(10), 822-830 (1980-10-01)
Sulfobetaines (N-alkyl-N,N-dimethyl-3-ammonio-1-propanesulfonates) have been identified as relatively specific and selective inhibitors of mitochondrial carnitine-acylcarnitine translocase. Thus, sublytic concentrations of sulfobetaines (alkyl = octyl to tetradecyl) inhibit the respiration of rat heart mitochondria supported by added acylcarnitines or pyruvate plus malonate
Xuecong Pan et al.
Langmuir : the ACS journal of surfaces and colloids, 34(18), 5273-5278 (2018-04-20)
Cooperative effects of a series of equimolar binary zwitterionic-ionic surfactant mixtures on the interfacial water structure at the air-water interfaces have been studied by sum frequency generation vibrational spectroscopy (SFG-VS). For zwitterionic surfactant palmityl sulfobetaine (SNC16), anionic surfactant sodium hexadecyl

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service