Skip to Content
Merck
All Photos(2)

Key Documents

A1346

Sigma-Aldrich

ASB-14

Synonym(s):

3-[N,N-Dimethyl(3-myristoylaminopropyl)ammonio]propanesulfonate, Amidosulfobetaine-14

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H46N2O4S
CAS Number:
Molecular Weight:
434.68
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.28
Pricing and availability is not currently available.

description

zwitterionic

Quality Level

Assay

≥95.0% (HPLC)

form

powder

mol wt

average mol wt >10,000

aggregation number

≥23

storage temp.

2-8°C

SMILES string

[S](=O)(=O)([O-])CCC[N+](CCCNC(=O)CCCCCCCCCCCCC)(C)C

InChI

1S/C22H46N2O4S/c1-4-5-6-7-8-9-10-11-12-13-14-17-22(25)23-18-15-19-24(2,3)20-16-21-29(26,27)28/h4-21H2,1-3H3,(H-,23,25,26,27,28)

InChI key

UTSXERRKRAEDOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

ASB-14 comprises alkyl tails and amidosulfobetaine headgroup.[1]

Application

ASB-14 has been used as a component of rehydration buffer for proteomic sample preparation from Leishmania donovani.[2]
Zwitterionic detergent. Useful for solubilization of proteins, including membrane proteins, for 2D electrophoresis.

Biochem/physiol Actions

ASB-14 is useful in proteasome extraction especially for the membrane associated proteins.[3] Along with CHAPS (3-(3-cholamidopropyl)dimethylammonio)-1-propanesulfonate), it is employed in protein solubilization even in the presence of thiourea.[4]

Analysis Note

ASB-14 (amidosulfobetaine-14), a membrane solubilizing zwitterionic detergent (medium chain amidosulfobetaine surfactant), may be used in protein and macromolecule research for the preparative extraction and solubilization of proteins for separation and analysis by methods such as 2D-electrophoresis. ASB-14 may be used in tissue engineering for antigen removal (AR) for xenogeneic scaffold generation. ASB-14 may be used to develop and study zwitterionic micelles.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protein Sample Preparation for Proteomic Analysis in Leishmania donovani
Alexandros A and Despina S
Bio-protocol, 4(5), 70-75 (2014)
Francisella t ularensis Proteome: Low Levels of ASB-14 Facilitate the Visualization of Membrane Proteins in Total Protein Extracts
Twine SM, et al.
Journal of Proteome Research, 4(5), 1848-1854 (2005)
Nityakalyani Srinivas et al.
Science (New York, N.Y.), 327(5968), 1010-1013 (2010-02-20)
Antibiotics with new mechanisms of action are urgently required to combat the growing health threat posed by resistant pathogenic microorganisms. We synthesized a family of peptidomimetic antibiotics based on the antimicrobial peptide protegrin I. Several rounds of optimization gave a
Thermodynamic and structural characterization of zwitterionic micelles of the membrane protein solubilizing amidosulfobetaine surfactants ASB-14 and ASB-16
D Andrea MG, et al.
Langmuir, 27(13), 8248-8256 (2011)
Sergej Skvortsov et al.
Molecular cancer therapeutics, 6(6), 1898-1908 (2007-06-19)
This study aimed to characterize the antitumor activity of 5-Chloro-N-[2-[2-(4-chloro-phenyl)-3-methyl-butoxy]-5-trifluoromethyl-phenyl]-2-hydroxy-benzamide (CTFB), a novel anticancer agent, in head and neck cancer cell lines, FaDu, SCC-25 and cisplatin-resistant CAL-27. CTFB was generated as a result of an extensive medicinal chemistry effort on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service