Skip to Content
Merck
All Photos(1)

Documents

H5257

Sigma-Aldrich

Hispidin

solid, ≥98% (HPLC)

Synonym(s):

6-(3,4-dihydroxystyrl)-4-hydroxy-2-pyrone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10O5
CAS Number:
Molecular Weight:
246.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

protect from light

color

yellow to brown

mp

237.5-238.5  °C

solubility

DMSO: >10 mg/mL

storage temp.

−20°C

SMILES string

OC1=CC(/C=C/C(O2)=CC(O)=CC2=O)=CC=C1O

InChI

1S/C13H10O5/c14-9-6-10(18-13(17)7-9)3-1-8-2-4-11(15)12(16)5-8/h1-7,14-16H/b3-1+

InChI key

SGJNQVTUYXCBKH-HNQUOIGGSA-N

Gene Information

General description

Hispidin is a phenolic compound, that is obtained from a medicinal mushroom, Phellinus linteus.

Biochem/physiol Actions

Hispidin exhibits robust antioxidant, anticancer and antidiabetic properties. It has the ability to guard against peroxynitrite-mediated cytotoxicity, DNA damage and the development of hydroxyl radicals.
Potent inhibitor of protein kinase Cβ, cytotoxic for cancer cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alvaro Macias et al.
International journal of molecular sciences, 22(3) (2021-02-13)
KV1.5 channel function is modified by different regulatory subunits. KVβ1.3 subunits assemble with KV1.5 channels and induce a fast and incomplete inactivation. Inhibition of PKC abolishes the KVβ1.3-induced fast inactivation, decreases the amplitude of the current KV1.5-KVβ1.3 and modifies their
Sirosh M Bokhari et al.
The Journal of investigative dermatology, 126(2), 460-467 (2005-12-24)
Activation of protein kinase C (PKC) induces phenotypic changes in the morphology of microvascular endothelial cells that affect major functions of the microvasculature. These functions include the first stages of sprouting in angiogenesis, cell migration following wounding, and vascular permeability.
Prabodh Risal et al.
Journal of natural products, 75(10), 1683-1689 (2012-10-03)
In this study the protective effects of davallialactone (1), isolated from Inonotus xeranticus, have been examined against carbon tetrachloride (CCl₄-induced acute liver injury. Mice received subcutaneous injection of 1 (2.5, 5, and 10 mg/kg) for three days before CCl₄ injection
Jinxi Huo et al.
Journal of advanced research, 24, 325-335 (2020-05-27)
Phellinus gilvus (Schwein.) Pat, a species of 'Sanghuang', has been well-documented for various medicinal uses, but the genome information and active constituents are largely unknown. Here, we sequenced the whole-genome of P. gilvus, identified phenylpropanoids as its key anti-cancer components
Craig Slattery et al.
The international journal of biochemistry & cell biology, 40(10), 2218-2229 (2008-04-29)
Cyclosporine A (CsA) significantly improves the success of organ transplantation, however renal fibrosis, characterised by severe tubulointerstitial fibrosis is a complication of CsA therapy. Previously we have reported the involvement of PKC-beta isoforms in a model of CsA-induced tubulointerstitial fibrosis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service