Skip to Content
Merck
All Photos(2)

Key Documents

H2775

Sigma-Aldrich

DL-threo-β-Hydroxyaspartic acid

Synonym(s):

threo-2-Amino-3-hydroxysuccinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7NO5
CAS Number:
Molecular Weight:
149.10
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.32

Quality Level

storage temp.

−20°C

SMILES string

N[C@H]([C@@H](O)C(O)=O)C(O)=O

InChI

1S/C4H7NO5/c5-1(3(7)8)2(6)4(9)10/h1-2,6H,5H2,(H,7,8)(H,9,10)/t1-,2-/m1/s1

InChI key

YYLQUHNPNCGKJQ-JCYAYHJZSA-N

Gene Information

human ... GLUL(2752)
mouse ... GLUL(14645)

Looking for similar products? Visit Product Comparison Guide

General description

DL-threo-β-hydroxyaspartic acid (THA) is a glutamate uptake inhibitor.

Application

DL-threo-β-Hydroxyaspartic acid (THA) has been used to block glutamate transport in cannulated sprague dawley rat.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jixu Yu et al.
Brain research, 1257, 102-107 (2009-01-13)
Threohydroxyaspartate (THA)-induced glutamate excitotoxicity in organotypic culture of rat spinal cord is a well-known model of motor neuron degeneration. THA causes accumulation of synaptic glutamate and over stimulation of the postsynaptic receptor by inhibiting glutamate uptake. This model has also
Richelle A Hemendinger et al.
Neurotoxicity research, 13(1), 49-61 (2008-03-28)
Amyotrophic lateral sclerosis (ALS) is a neurodegenerative disease resulting from the progressive loss of motor neurons in the spinal cord and brain. To date, clinically effective neuroprotective agents have not been available. The current study demonstrates for the first time
Yasuhiro Kosuge et al.
Neuroscience letters, 454(2), 165-169 (2009-05-12)
Amyotrophic lateral sclerosis (ALS) is a devastating neurodegenerative disease characterized by selective loss of motor neurons. Although organotypic spinal slice cultures (OSCs) exposed to inhibitors of glutamate uptake have been used as a model of ALS for screening of potentially
Z Kawakami et al.
Neuroscience, 159(4), 1397-1407 (2009-05-05)
To clarify the mechanism of yokukansan (TJ-54), a traditional Japanese medicine, against glutamate-mediated excitotoxicity, the effects of TJ-54 on glutamate uptake function were first examined using cultured rat cortical astrocytes. Under thiamine-deficient conditions, the uptake of glutamate into astrocytes, and
Non-heme hydroxylase engineering for simple enzymatic synthesis of L-threo-hydroxyaspartic acid.
Matthias Strieker et al.
Chembiochem : a European journal of chemical biology, 9(3), 374-376 (2008-01-24)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service