Skip to Content
Merck
All Photos(3)

Key Documents

I8896

Sigma-Aldrich

IGEPAL® CA-630

Synonym(s):

Octylphenoxy poly(ethyleneoxy)ethanol, branched

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H4O)nC14H22O
CAS Number:
UNSPSC Code:
12352104
NACRES:
NA.31

grade

Molecular Biology

Quality Level

description

non-ionic

form

liquid

technique(s)

IR spectroscopy: suitable

CMC

0.08 mM (20-25°C)

HLB

13

foreign activity

DNase and RNase, none detected

SMILES string

O(CCO)c1ccc(cc1)C(CC(C)(C)C)(C)C

InChI

1S/C16H26O2/c1-15(2,3)12-16(4,5)13-6-8-14(9-7-13)18-11-10-17/h6-9,17H,10-12H2,1-5H3

InChI key

JYCQQPHGFMYQCF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

IGEPAL® CA-630 is a nonionic, non-denaturing detergent suitable for solubilization, isolation and purification of membrane protein complexes.It is chemically indistinguishable from Nonidet P-40.

Application

IGEPAL® CA-630 has been used
  • as neutral detergent in homogenizing buffer during western blotting of mouse muscle homogenates
  • In X-gal staining for studying the lacZ expression in E8.5-E10.5 embryos
  • In Cell-Lysis (CL) buffer for preparing RT-qPCR-ready cell lysates

Other Notes

Nonionic, non-denaturing detergent

Quality

Chemically indistinguishable from Nonidet P-40, which is no longer commercially available.

Caution

May develop turbidity or sediment during storage. A clear liquid can be obtained on heating to 40°C.

Legal Information

IGEPAL is a registered trademark of Solvay

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

483.8 °F - closed cup

Flash Point(C)

251 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Joseph A Roche et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 59(11), 964-975 (2011-11-02)
Mutations in the DYSF gene that severely reduce the levels of the protein dysferlin are implicated in muscle-wasting syndromes known as dysferlinopathies. Although studies of its function in skeletal muscle have focused on its potential role in repairing the plasma
Bojana Lucic et al.
Nature communications, 10(1), 4059-4059 (2019-09-08)
HIV-1 recurrently targets active genes and integrates in the proximity of the nuclear pore compartment in CD4+ T cells. However, the genomic features of these genes and the relevance of their transcriptional activity for HIV-1 integration have so far remained
Amina Abbadi et al.
The Journal of biological chemistry, 291(3), 1448-1455 (2015-11-26)
Many cells, including murine airway epithelial cells, respond to a variety of inflammatory stimuli by synthesizing leukocyte-adhesive hyaluronan (HA) cables that remain attached to their cell surfaces. This study shows that air-liquid interface cultures of murine airway epithelial cells (AECs)
Per Wasteson et al.
Development (Cambridge, England), 135(10), 1823-1832 (2008-04-18)
Aortic smooth muscle cells (SMCs) have been proposed to derive from lateral plate mesoderm. It has further been suggested that induction of SMC differentiation is confined to the ventral side of the aorta, and that SMCs later migrate to the
Simon Lalonde et al.
Genome biology, 20(1), 133-133 (2019-07-10)
Genome-wide association studies (GWAS) have identified hundreds of loci associated with coronary artery disease (CAD) and blood pressure (BP) or hypertension. Many of these loci are not linked to traditional risk factors, nor do they include obvious candidate genes, complicating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service