H1404
Hepatocyte Growth Factor human
>95% (SDS-PAGE), recombinant, expressed in Baculovirus infected High-5 cells, lyophilized powder, suitable for cell culture
Synonym(s):
HGF, Hepatopoietin A, Scatter Factor
About This Item
Recommended Products
product name
Hepatocyte Growth Factor human, HGF, recombinant, expressed in Baculovirus infected High-5 cells, suitable for cell culture
biological source
human
Quality Level
recombinant
expressed in Baculovirus infected High-5 cells
Assay
>95% (SDS-PAGE)
form
lyophilized powder
potency
1.0-20.0 ng/mL
quality
endotoxin tested
mol wt
~80 kDa
packaging
pkg of 5x5 μg
pkg of 5 μg
storage condition
avoid repeated freeze/thaw cycles
technique(s)
cell culture | mammalian: suitable
impurities
<1.0 EU/μg
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... HGF(3082)
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Other Notes
Physical form
Preparation Note
Analysis Note
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service