Skip to Content
Merck
All Photos(1)

Documents

H0879

Sigma-Aldrich

Hexamethonium bromide

Synonym(s):

Hexane-1,6-bis(trimethylammonium bromide), N,N,N,N′,N′,N′-Hexamethylhexamethylenediammonium dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N(Br)(CH2)6N(Br)(CH3)3
CAS Number:
Molecular Weight:
362.19
Beilstein:
3715749
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

mp

282-285 °C (dec.)

SMILES string

[Br-].[Br-].C[N+](C)(C)CCCCCC[N+](C)(C)C

InChI

1S/C12H30N2.2BrH/c1-13(2,3)11-9-7-8-10-12-14(4,5)6;;/h7-12H2,1-6H3;2*1H/q+2;;/p-2

InChI key

FAPSXSAPXXJTOU-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Hexamethonium bromide has been used:
  • as an autonomic ganglia-blocking agent to study its effects on whisker movement in rats
  • as a nicotinic receptor antagonist to study its effects on antigen-induced arthritis (AIA) progression in mice
  • as a non-selective ganglionic nicotinic acetylcholine receptor (nAChR) antagonist to study its effects on cytisine-induced autonomic cardiovascular responses in mice

Biochem/physiol Actions

Hexamethonium bromide is a nicotinic acetylcholine receptor (nAChR) antagonist. It preferentially blocks nicotinic receptors at autonomic ganglia. It can cross the blood-brain barrier only at high doses.

Features and Benefits

This compound is featured on the Acetylcholine Receptors (Nicotinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piyali Dasgupta et al.
Proceedings of the National Academy of Sciences of the United States of America, 103(16), 6332-6337 (2006-04-08)
Non-small cell lung cancer (NSCLC) demonstrates a strong etiologic association with smoking. Although nicotine is not carcinogenic, it can induce cell proliferation and angiogenesis and suppress apoptosis induced by certain agents. Here we show that nicotine inhibits apoptosis induced by
James T Heaton et al.
Neuroscience, 265, 9-20 (2014-02-01)
Vibrissal whisking is often employed to track facial nerve regeneration in rats; however, we have observed similar degrees of whisking recovery after facial nerve transection with or without repair. We hypothesized that the source of non-facial nerve-mediated whisker movement after
Alexandre Kanashiro et al.
Basic & clinical pharmacology & toxicology, 119(5), 453-457 (2016-04-22)
The cholinergic anti-inflammatory pathway (CAP) is a complex neuroimmune mechanism triggered by the central nervous system to regulate peripheral inflammatory responses. Understanding the role of CAP in the pathogenesis of rheumatoid arthritis (RA) could help develop new therapeutic strategies for
Simon Akerman et al.
Brain : a journal of neurology, 135(Pt 12), 3664-3675 (2012-10-16)
Trigeminal autonomic cephalalgias are highly disabling primary headache disorders, characterized by severe unilateral head pain and associated ipsilateral cranial autonomic features. There is limited understanding of their pathophysiology and how and where treatments act to reduce symptoms; this is significantly
Hiroshi Hanamoto et al.
International journal of oral science, 4(1), 24-29 (2012-03-06)
Stimulation of the trigeminal nerve can elicit various cardiovascular and autonomic responses; however, the effects of anesthesia with pentobarbital sodium on these responses are unclear. Pentobarbital sodium was infused intravenously at a nominal rate and the lingual nerve was electrically

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service