Skip to Content
Merck
All Photos(2)

Key Documents

H0878

Sigma-Aldrich

Heparin lithium salt from porcine intestinal mucosa

≥150 USP units/mg

Synonym(s):

Lithium acid heparin, Lithium heparinate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

Porcine intestinal mucosa

Quality Level

form

powder

specific activity

≥150 USP units/mg

color

off-white

solubility

water: 40 mg/mL, clear, colorless to faint yellow or tan

cation traces

Li: 3-4%

application(s)

diagnostic assay manufacturing
life science and biopharma
plasma purification
sample preparation

storage temp.

room temp

SMILES string

[S](=O)(=O)(NC1C(OC(C(C1O)OC2OC(C(C(C2O[S](=O)(=O)O)O)OC)C(=O)O)CO[S](=O)(=O)O)OC)O

InChI

1S/C14H25NO20S3/c1-29-9-7(17)10(35-38(26,27)28)14(34-11(9)12(18)19)33-8-4(3-31-37(23,24)25)32-13(30-2)5(6(8)16)15-36(20,21)22/h4-11,13-17H,3H2,1-2H3,(H,18,19)(H,20,21,22)(H,23,24,25)(H,26,27,28)

InChI key

HXSDFQWQRCUQHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Heparin lithium is a highly sulfated polysaccharide that belongs to the family of glycosaminoglycans with remarkable significance in the areas of cell culture, cancer biology, biochemical, and drug discovery research. Due to its interactions with proteins and cells, it is used to study molecular and cellular processes, tumor modeling, investigate binding interactions, and as a cell culture supplement, to maintain cell viability.

Application

Heparin lithium was used:
  • To validate photometric measurements in cell surface charge determination
  • As an anti-coagulant for laboratory studies
  • To study hypothermal stress on lactate metabolism
  • To study the effects of anticoagulants in direct membrane feeding assays
  • For 2D and 3D vitro cell culture in glioblastoma cell models
  • To prevent clot formation and tissue damage during mechanical probing of cardiac pacemaker cell embryonic patterning, a study that contributes to the broader area of cardiac tissue mechanics research and developmental biology

Features and Benefits

  • Heparin with specific activity of ≥150 USP units/mg
  • Versatile and adaptable for a wide variety of laboratory and research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.
To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of anticoagulants in blood collection tubes on Plasmodium falciparum transmission in direct membrane feeding assays
Wouter G et al.
Transactions of the Royal Society of Tropical Medicine and Hygiene, 116, 187- 189 (2022)
Determination of cell surface charge by photometric titration
Thethi K et al
Journal of Biochemical and Biophysical Methods, 34, 137-145 (1997)
Chia-Hao Chang et al.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology, 248, 110744-110744 (2020-06-23)
The milkfish Chanos chanos, an economically important cultured marine species in Southeast Asia, exhibits stenothermal and euryhaline characteristics and huge mortality usually occurs during extreme cold weather in winter. Under conditions beyond optimal temperatures, ectothermic species experience an increase in
Beatriz Giesen et al.
Pharmaceutics, 13(2) (2021-03-07)
Gold nanoparticles (Au NPs) are studied as delivery systems to enhance the effect of the glutaminase1 inhibitor CB839, a promising drug candidate already in clinical trials for tumor treatments. Au NPs were synthesized using a bottom-up approach and covered with
R B Shartau et al.
The Journal of experimental biology, 223(Pt 7) (2020-03-05)
Acute (<96 h) exposure to elevated environmental CO2 (hypercarbia) induces a pH disturbance in fishes that is often compensated by concurrent recovery of intracellular and extracellular pH (pHi and pHe, respectively; coupled pH regulation). However, coupled pH regulation may be limited

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service