Skip to Content
Merck
All Photos(1)

Key Documents

E0270

Sigma-Aldrich

Ethylenediaminetetraacetic acid tripotassium salt dihydrate

98% (titration)

Synonym(s):

EDTA tripotassium salt, Edathamil, Tripotassium ethylenediaminetetraacetate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(KOOCCH2)2NCH2CH2N(CH2COOK)CH2COOH · 2H2O
CAS Number:
Molecular Weight:
442.54
Beilstein:
6762497
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.31

biological source

synthetic

Quality Level

Assay

98% (titration)

form

powder

reaction suitability

reagent type: chelator

pH

8-9 (20 °C, 50 g/L)

mp

182 °C (dec.) (lit.)

solubility

3 M KOH: 160 mg/mL, clear, colorless

storage temp.

room temp

SMILES string

[K+].[K+].[K+].[H]O[H].[H]O[H].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.3K.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;;2*1H2/q;3*+1;;/p-3

InChI key

MAPFUJCWRWFQIY-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Ethylenediaminetetraacetic acid tripotassium salt dihydrate has been used to coat conical tubes to collect trunk blood for tissue and plasma samples. It has also been used in tubes to collect blood to perform mixed meal test (MMT).
Used to eliminate inhibition of enzyme catalyzed reactions due to traces of heavy metals

Biochem/physiol Actions

Ethylene diamine tetraacetic acid (EDTA) chelates divalent cations like calcium and magnesium in whole blood. It is usually available as a salt, complexed with either potassium or sodium. It is mostly used during hematological tests of the blood sample. It helps to maintain the cellular morphology to prepare blood smears for microscopic analysis. EDTA is abundantly used in RNA purification methods.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

EPR methods to study specific metal-ion binding sites in RNA
Test, 468, 335-367 (2009)
Blood Sample Collection and Handling
Comprehensive Sampling and Sample Preparation (2012)
Post-surgical effects of roux-En-Y gastric bypass on glucose homeostasis, intestinal morphology and L-cells in obese Gottingen minipigs
Barkholt P, et al.
Journal of Obesity and Bariatrics, 1(1), 8-8 (2015)
Nathan C Bingham et al.
Endocrinology, 149(5), 2138-2148 (2008-02-09)
Leptin, an adipocyte-derived hormone, has emerged as a critical regulator of energy homeostasis. The leptin receptor (Lepr) is expressed in discrete regions of the brain; among the sites of highest expression are several mediobasal hypothalamic nuclei known to play a
EPR methods to study specific metal-ion binding sites in RNA
Methods in Enzymology, 468, 335-367 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service