Skip to Content
Merck
All Photos(2)

Key Documents

GHS1128

Sigma-Aldrich

Hematoxylin Solution, Gill No. 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116124
PubChem Substance ID:
NACRES:
NA.47

form

solution

Quality Level

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

2 g/L

pH

2.5-3.5

application(s)

hematology
histology

storage temp.

room temp

SMILES string

Oc1cc2C[C@@]3(O)COc4c(O)c(O)ccc4[C@H]3c2cc1O

InChI

1S/C16H14O6/c17-10-2-1-8-13-9-4-12(19)11(18)3-7(9)5-16(13,21)6-22-15(8)14(10)20/h1-4,13,17-21H,5-6H2/t13-,16+/m0/s1

InChI key

WZUVPPKBWHMQCE-XJKSGUPXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Gill No. 1 formulation is used as a progressive cytology stain. Used with hematoxylin and eosin staining.

Other Notes

2 g/L certified hematoxylin

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael J Turner et al.
Journal of neuroimmunology, 285, 4-12 (2015-07-23)
Alemtuzumab, a monoclonal antibody directed against human CD52, is used in the treatment of MS. To characterize the impact of anti-CD52 administration, a monoclonal antibody to mouse CD52 (anti-muCD52) was generated and evaluated in EAE mouse models of MS. A
Eleni Maniati et al.
Cell reports, 30(2), 525-540 (2020-01-16)
Although there are many prospective targets in the tumor microenvironment (TME) of high-grade serous ovarian cancer (HGSOC), pre-clinical testing is challenging, especially as there is limited information on the murine TME. Here, we characterize the TME of six orthotopic, transplantable syngeneic
Hideki Ishii et al.
Bioorganic & medicinal chemistry letters, 22(3), 1469-1474 (2012-01-17)
SAR studies for the exploration a novel class of anti-human immunodeficiency virus type 1 (HIV-1) agents based on the hematoxylin structure (1) are described. The systematic deoxygenations of 1 including asymmetric synthesis were conducted to obtain a compound showing high
Li-Gen Lin et al.
Journal of medicinal chemistry, 51(15), 4419-4429 (2008-07-10)
Protein tyrosine kinase (PTK) inhibitors represent emerging therapeutics for cancer chemoprevention. In our study, hematoxylin (26) was identified as one of the most remarkable c-Src inhibitors in an orthogonal compound-mixing library (32200 compounds) by using an ELISA-based automated high-throughput screening
Devalingam Mahalingam et al.
Autophagy, 10(8), 1403-1414 (2014-07-06)
We previously reported that inhibition of autophagy significantly augmented the anticancer activity of the histone deacetylase (HDAC) inhibitor vorinostat (VOR) through a cathepsin D-mediated mechanism. We thus conducted a first-in-human study to investigate the safety, preliminary efficacy, pharmacokinetics (PK), and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service