Skip to Content
Merck
All Photos(1)

Key Documents

GEE70092Z

Shrimp Alkaline Phosphatase

Cytiva E70092Z, pack of 1000 U

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.54

form

solid

packaging

pack of 1000 U

manufacturer/tradename

Cytiva E70092Z

shipped in

dry ice

storage temp.

−20°C

General description

Shrimp alkaline phosphatase is a high specific activity, heat-labile alkaline phosphatase. Completely and irreversibly inactivated in Tris buffers at pH 8.0 to 8.5 by heating for 15 min at 65°C. No further treatment is necessary. The ALP (alkaline phosphatase) gene codes for a membrane-bound ectoenzymes that is present in four isoforms and is distributed as tissue nonspecific, intestinal, placenta, and germ cell ALP. The tissue nonspecific enzyme is ubiquitously expressed in a variety of tissues including liver, bone, kidney and is predominant in osteoblasts. Alkaline phosphatase is also known as mesenchymal stem cell antigen 1 or tissue nonspecific alkaline phosphatase.

Biochem/physiol Actions

Alkaline phosphatase hydrolyses pyrophosphate (PPi) to inorganic phosphate and thus maintains a balance between the two forms of phosphate which is highly essential for mineralization. Higher levels of serum alkaline phosphatase is found in prostate cancer. Severe TNAP deficiency refers to hypophosphatasia characterized with defective pyridoxal 5′-phosphate metabolism and skeletal hypomineralization.

Features and Benefits

  • Removing 5′-phosphates from DNA and RNA.
  • Easily inactivated by heat.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tumour-derived alkaline phosphatase regulates tumour growth, epithelial plasticity and disease-free survival in metastatic prostate cancer.
Rao S R, et al.
British Journal of Cancer, 116(2), 227-227 (2017)
The mechanism of mineralization and the role of alkaline phosphatase in health and disease.
Orimo H
Journal of Nippon Medical School = Nippon Ika Daigaku Zasshi, 77(1), 4-12 (2010)
Increased activity of TNAP compensates for reduced adenosine production and promotes ectopic calcification in the genetic disease ACDC.
Jin H, et al.
Science Signaling, 9(458), ra121-ra121 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service