GE27-4563-01
Calf Thymus Sonicated DNA
Cytiva 27-4563-01
About This Item
Recommended Products
packaging
pkg of 500 μg/mL
manufacturer/tradename
Cytiva 27-4563-01
shipped in
dry ice
storage temp.
−20°C
Related Categories
General description
- Tested for absence of nuclease.
- Fragmented DNA with a size range from 200-6000 nucleotides.
- Purified by phenol extraction and freeze dried.
- Suitable for blocking non-specific binding sites in hybridization experiments to reduce background.
DNA isolated from Calf thymus is sonicated to fragment the DNA to an average size of approximately 400 bp.
Storage and Stability
Analysis Note
related product
Storage Class Code
13 - Non Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service