Skip to Content
Merck
All Photos(1)

Key Documents

D3664

Sigma-Aldrich

Deoxyribonucleic acid sodium salt from calf thymus

suitable for substrate for DNase (Readily soluble)

Synonym(s):

CT-DNA, calf thymus DNA, DNA sodium salt from calf thymus, Thymonucleic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106305
eCl@ss:
32160414
NACRES:
NA.51

form

powder or crystals

suitability

suitable for substrate for DNase (Readily soluble)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Calf thymus DNA (CT-DNA) is a natural DNA widely used in studies of DNA binding anticancer agents and DNA binding agents that modulate DNA structure and function. Calf thymus DNA is also used in physicochemical studies of DNA behavior in solution.

Packaging

Preweighed vials prepared from D1501

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristin Grøsvik et al.
Frontiers in microbiology, 11, 263-263 (2020-03-12)
The cellular methyl donor S-adenosylmethionine (SAM) and other endo/exogenous agents methylate DNA bases non-enzymatically into products interfering with replication and transcription. An important product is 3-methyladenine (m3A), which in Escherichia coli is removed by m3A-DNA glycosylase I (Tag) and II
Abdollah Zakeri Siavashani et al.
Biomaterials science, 8(1), 148-162 (2019-10-31)
Implantation of temporary and permanent biomaterials in the body leads to a foreign body reaction (FBR), which may adversely affect tissue repair processes and functional integration of the biomaterial. However, modulation of the inflammatory response towards biomaterials can potentially enable
Xingyi Chen et al.
Molecular pharmaceutics, 16(6), 2690-2699 (2019-05-01)
The nucleotide-binding domain and leucine-rich repeat-containing family, pyrin domain-containing 3 (NLRP3) inflammasome is a key regulator of innate immune responses, and its aberrant activation is implicated in the pathogenesis of many diseases such as Alzheimer's disease and type 2 diabetes.
Coordination ability and biological activity of a naringenin thiosemicarbazone.
Katarzyna Brodowska et al.
Journal of inorganic biochemistry, 165, 36-48 (2016-10-21)
Farhad Ahmadi et al.
DNA and cell biology, 31(2), 259-268 (2011-08-25)
In this work, multispectroscopic atomic force microscopy and molecular modeling [ONIOM 2(B3LYP/6-31++G(d,p): Universal Force Field (UFF)) level] techniques were used to study the interaction between Calf-Thymus-DNA (CT-DNA) and pyriproxyfen (PYR) insecticide. The binding constant of PYR with double-strand deoxyribonucleic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service