GE17-5157-01
HiTrap® SP Fast Flow
Cytiva 17-5157-01, pack of 5 × 5 mL
Synonym(s):
HiTrap SP
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
23151816
NACRES:
NA.56
Recommended Products
ligand
sulphopropyl
description
Ion Exchanger Type (value)
packaging
pack of 5 × 5 mL
manufacturer/tradename
Cytiva 17-5157-01
parameter
flow rate
42 psi
bed size
16 mm × 25 mm
bed volume
5 mL
column I.D.
16 mm
matrix
6% cross-linked agarose
particle size
45-165 μm
average diameter
90 μm
cleaning
3-14
working range
4-13
suitability
suitable for bioprocess medium
General description
HiTrap® SP FF is prepacked with SP Sepharose™ Fast Flow, and is a strong cation exchanger for small-scale protein purifications as well as screening of binding and elutions conditions.
Q, SP, DEAE, and CM Sepharose™ Fast Flow are based on a robust, 6% highly cross-linked beaded agarose matrix with good flow properties and high loading capacities. ANX Sepharose™ 4 Fast Flow (high sub) is based on 4% highly cross-linked beaded agarose. This results in a medium with higher porosity, which is particularly useful for the purification of high molecular mass proteins.
Q, SP, DEAE, and CM Sepharose™ Fast Flow are based on a robust, 6% highly cross-linked beaded agarose matrix with good flow properties and high loading capacities. ANX Sepharose™ 4 Fast Flow (high sub) is based on 4% highly cross-linked beaded agarose. This results in a medium with higher porosity, which is particularly useful for the purification of high molecular mass proteins.
Features and Benefits
- Convenient and affordable for fast, easy ion exchange separations either alone or connected in series.
- The industry standard for ion exchange chromatography.
- High flow rates and good scale-up potential.
- Use a weak ion exchanger if the selectivity of a strong ion exchanger is insufficient.
- Predictable scale-up
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (0.2 M sodium acetate in 20% ethanol)
Store at 4 to 30 °C (20% Ethanol + 0.2 M Sodium Acetate)
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Other Notes
The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.
Legal Information
HiTrap is a registered trademark of Cytiva
Sepharose is a trademark of Cytiva
related product
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service