GE17-0440-03
Con-A Sepharose™ 4B
Cytiva 17-0440-03, pack of 5 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106500
NACRES:
NA.56
Recommended Products
ligand
concanavalin A (con A)
packaging
pack of 5 mL
manufacturer/tradename
Cytiva 17-0440-03
storage condition
(0.1 M acetate buffer pH 6, 1 M NaCl, 1 mM CaCl2, 1 mM MnCl2 and 1 mM MgCl, 20% Ethanol)
matrix
4% agarose
particle size
45-165 μm
average diameter
90 μm
cleaning in place
4-9
working range
4-6
storage temp.
2-8°C
Related Categories
General description
Con A Sepharose™ 4B with immobilized Concanavalin A is an affinity chromatography media for capture of glycosylated biomolecules including glycoproteins and glycolipids. Stronger affinity for α-D-glucose than α-D-mannose.
Application
Con A Sepharose™ is Concanavalin A coupled to Sepharose™ 4B by the cyanogen bromide method. Concanavalin A (Con A) is a tetrameric metalloProtein isolated from Canavalia ensiformis (jack bean). Con A binds molcules containing α-D-mannopyranosyl, α-D-glucopyranosyl and sterically related residues. The binding sugar requires the presence of C-3, C-4 and C-5 hydroxyl groups for reaction with Con A. Con A coupled to Sepharose™ is routinely used for separation and purification of glycoProteins, polysaccharides and glycolipids
Features and Benefits
- Concanavalin A (Con A) binds molecules that contain α-D-mannose, α-D-glucose, and sterically related residues with available C-3, C-4, or C-5 hydroxyl groups
- Group-specific adsorbent for molecules containing sugars.
- Highly suitable for isolation of cell surface glycoProteins from detergent-solubilized membranes
- Concanavalin A (Con A) binds molecules that contain α-D-mannose, α-D-glucose and sterically related residues with available C-3, C-4, or C-5 hydroxyl groups
- Highly suitable for isolation of cell surface glycoProteins from detergent-solubilized membranes
- Con A coupled to Sepharose™ 4B via cyanogen bromide activation
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
Sepharose is a trademark of Cytiva
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service