Skip to Content
Merck
All Photos(2)

Key Documents

67770

Millipore

Methyl α-D-mannopyranoside

suitable for microbiology, ≥99.0%, enables differentiation between species of Listeria

Synonym(s):

Methyl alpha-D-mannoside, Methyl-alpha-D-mannopyranoside, α-Methyl D-mannoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14O6
CAS Number:
Molecular Weight:
194.18
Beilstein:
81566
EC Number:
MDL number:
UNSPSC Code:
41106212
PubChem Substance ID:
NACRES:
NA.85

Quality Level

Assay

≥99.0% (sum of enantiomers, HPLC)
≥99.0%

form

powder

optical activity

[α]20/D 77.0 to 82.0°, c = 10% in H2O

mol wt

194.18 g/mol

mp

187-195 °C
193-196 °C (lit.)

solubility

H2O: 0.1 g/mL, clear, colorless

application(s)

microbiology

SMILES string

CO[C@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@@H]1O

InChI

1S/C7H14O6/c1-12-7-6(11)5(10)4(9)3(2-8)13-7/h3-11H,2H2,1H3/t3-,4-,5+,6+,7+/m1/s1

InChI key

HOVAGTYPODGVJG-VEIUFWFVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl α-D-mannopyranoside is a compound that belongs to the class of organic compounds known as o-glycosyl compounds. It is used for the differentiation of Listeria species, Listeria monocytogenes, Listeria innocua, and Listeria welshimeri can ferment the sugar, producing acid which can be identified using an appropriate pH indicator. It has been used to synthesize a series of tri- and tetrahydroxylated seven-membered imino sugars in a study that worked towards a stable Noeuromycin (glycosyl cation mimic that strongly inhibits glycosidases) analog with a D-manno configuration. It has also been used in a study to investigate the primary mannose-binding site of Pradimicin A. (antifungal agent)

Application

Methyl α-D-mannopyranoside can be used to identify different species of Listeria based on their ability to ferment the sugar.

Other Notes

Unwanted binding of avidin to endogenous lectins

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

404.1 °F

Flash Point(C)

206.74 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prevention of nonspecific binding of avidin.
R C Duhamel et al.
Methods in enzymology, 184, 201-207 (1990-01-01)
Oliver Schwardt et al.
Bioorganic & medicinal chemistry, 19(21), 6454-6473 (2011-10-04)
Urinary tract infection (UTI) caused by uropathogenic Escherichia coli (UPEC) is one of the most prevalent infectious diseases. Particularly affected are women, who have a 40-50% risk to experience at least one symptomatic UTI episode at some time during their
Karen T Welch et al.
Bioorganic & medicinal chemistry letters, 18(24), 6573-6575 (2008-11-08)
A virtual screening approach was used to identify new glycomimetics. The National Cancer Institute Diversity Set was docked into the carbohydrate binding site of the lectin concanavalin A (ConA). The resulting poses were analyzed and 19 molecules were tested for
Rafael Maldonado-Hernández et al.
Analytical biochemistry, 610, 113887-113887 (2020-08-09)
Over the past 10 years we have been developing a multi-attribute analytical platform that allows for the preparation of milligram amounts of functional, high-pure, and stable Torpedo (muscle-type) nAChR detergent complexes for crystallization purpose. In the present work, we have
Su Yu et al.
BMC gastroenterology, 9, 58-58 (2009-07-25)
GP2 is the major membrane protein present in the pancreatic zymogen granule, and is cleaved and released into the pancreatic duct along with exocrine secretions. The function of GP2 is unknown. GP2's amino acid sequence is most similar to that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service