Skip to Content
Merck
All Photos(1)

Key Documents

G7423

Sigma-Aldrich

GSK3787

≥98% (HPLC), white to off-white, powder

Synonym(s):

4-Chloro-N-(2-{[5-trifluoromethyl)-2-pyridyl]sulfonyl}ethyl)benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12ClF3N2O3S
CAS Number:
Molecular Weight:
392.78
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

solubility

DMSO: ≥10 mg/mL

originator

GlaxoSmithKline

storage temp.

2-8°C

SMILES string

O=S(C1=CC=C(C=N1)C(F)(F)F)(CCNC(C2=CC=C(Cl)C=C2)=O)=O

InChI

1S/C15H12ClF3N2O3S/c16-12-4-1-10(2-5-12)14(22)20-7-8-25(23,24)13-6-3-11(9-21-13)15(17,18)19/h1-6,9H,7-8H2,(H,20,22)

InChI key

JFUIMTGOQCQTPF-UHFFFAOYSA-N

Application

GSK3787 has been used to inhibit the role of peroxisome proliferator-activated receptor-delta (PPARδ), during the pre-implantation period of bovine embryonic development.[1] It has also been used as a PPARδ-specific inhibitor in in vitro maturation (IVM) media to inhibit PPARδ.[2]

Biochem/physiol Actions

GSK3787 is an orally available selective irreversible Peroxisome Proliferator-Activated Receptor δ (PPARδ) antagonist (pIC50=6.6) with no measurable affinity for hPPARR or hPPARγ (pIC50<5). It acts by covalently modifying Cys249 within the ligand binding pocket, and has been shown to antagonize the induction of PPARδ-regulated target genes in skeletal muscle cells.
GSK3787 is known to exhibit pharmacokinetic properties.[3]

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lina Bai et al.
Acta biochimica et biophysica Sinica, 52(12), 1373-1381 (2020-11-25)
Exposure to adverse factors in utero may lead to adaptive changes in cardiac structure and metabolism, which increases the risk of chronic cardiovascular disease later in life. Studies showed that the angiotensin II type 1 receptor autoantibodies (AT1-AAs) are able
Stefan Kluge et al.
Biochimica et biophysica acta. Molecular and cell biology of lipids, 1866(4), 158875-158875 (2021-01-10)
The α-tocopherol-derived long-chain metabolite (α-LCM) α-13'-carboxychromanol (α-13'-COOH) is formed via enzymatic degradation of α-tocopherol (α-TOH) in the liver. In the last decade, α-13'-COOH has emerged as a new regulatory metabolite revealing more potent or even different effects compared with its
Yen-Chung Chen et al.
PloS one, 8(7), e69702-e69702 (2013-07-12)
We hypothesized that prostacyclin (PGI2) protects vascular smooth muscle cell (VSMC) against apoptosis and phenotypic switch through peroxisome proliferator-activated receptor-α (PPARα) activation and 14-3-3 upregulation. Here we showed that transfection of rat aortic VSMC, A-10, with PGI2-producing vectors, Ad-COPI, resulted
Barry G Shearer et al.
Journal of medicinal chemistry, 53(4), 1857-1861 (2010-02-05)
4-Chloro-N-(2-{[5-trifluoromethyl)-2-pyridyl]sulfonyl}ethyl)benzamide 3 (GSK3787) was identified as a potent and selective ligand for PPARdelta with good pharmacokinetic properties. A detailed binding study using mass spectral analysis confirmed covalent binding to Cys249 within the PPARdelta binding pocket. Gene expression studies showed that
Kenta Kuramoto et al.
Genes, 12(1) (2021-01-21)
Cancer stem cells (CSCs), having both self-renewal and tumorigenic capacity, utilize an energy metabolism system different from that of non-CSCs. Lipid droplets (LDs) are organelles that store neutral lipids, including triacylglycerol. Previous studies demonstrated that LDs are formed and store

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service