Skip to Content
Merck
All Photos(4)

Key Documents

G5884

Sigma-Aldrich

Guanosine 5′-triphosphate lithium salt

~95% (HPLC), powder

Synonym(s):

GTP lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N5O14P3 · xLi+
CAS Number:
Molecular Weight:
523.18 (free acid basis)
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

Assay

~95% (HPLC)

Quality Level

form

powder

color

white

solubility

H2O: 100 mg/mL

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Li+].NC1=Nc2c(ncn2[C@@H]3O[C@H](COP(O)(=O)OP(O)(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C(=O)N1

InChI

1S/C10H16N5O14P3.Li/c11-10-13-7-4(8(18)14-10)12-2-15(7)9-6(17)5(16)3(27-9)1-26-31(22,23)29-32(24,25)28-30(19,20)21;/h2-3,5-6,9,16-17H,1H2,(H,22,23)(H,24,25)(H2,19,20,21)(H3,11,13,14,18);/q;+1/p-1/t3-,5-,6-,9-;/m1./s1

InChI key

WDCLDKGIVWUKSE-GWTDSMLYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Guanosine 5′-triphosphate lithium salt has been used:
  • In ribosome activity assay.
  • In tubulin isolation and polymerization.
  • In the determination of cell viability.

Biochem/physiol Actions

GTP functions as a carrier of phosphates and pyrophosphates involved in channeling chemical energy into specific biosynthetic pathways. GTP activates the signal transducing G proteins which are involved in various cellular processes including proliferation, differentiation, and activation of several intracellular kinase cascades. Proliferation and apoptosis are regulated in part by the hydrolysis of GTP by small GTPases Ras and Rho. Another type of small GTPase, Rab, plays a role in the docking and fusion of vesicles and may also be involved in vesicle formation. In addition to its role in signal transduction, GTP also serves as an energy-rich precursor of mononucleotide units in the enzymatic biosynthesis of DNA and RNA.
Guanosine 5′-triphosphate serves as a substrate for guanosine 5′-triphosphate 3′ diphosphate diphosphatase, a hydrolyzing enzyme and guanosine triphosphate guanyltransferase.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the P2 Receptors: P2X Ion Channel Family page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Caution

Stability is much better than sodium salt.

Preparation Note

Prepared by enzymatic phosphorylation of 5′-GMP.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alcoholic liver disease and the mitochondrial ribosome: methods of analysis
Alcohol, 381-394 (2008)
Effect of quinones on microtubule polymerization: a link between oxidative stress and cytoskeletal alterations in Alzheimer's disease
Santa-Maia I, et al.
Biochimica et Biophysica Acta (BBA)-Molecular Basis of Disease, 1740(3), 472-480 (2005)
Enzyme (2005)
Adelaine K W Leung et al.
Journal of molecular biology, 402(1), 154-164 (2010-07-21)
RNA is known to perform diverse roles in the cell, often as ribonucleoprotein (RNP) particles. While the crystal structure of these RNP particles could provide crucial insights into their functions, crystallographic work on RNP complexes is often hampered by difficulties
Aida Menéndez-Méndez et al.
STAR protocols, 2(4), 100964-100964 (2021-11-30)
Low-density cell culture of the postnatal cerebellum, combined with live imaging and single-cell tracking, allows the behavior of postnatal cerebellar neural stem cells (NSCs) and their progeny to be monitored. Cultured cerebellar NSCs maintain their neurogenic nature giving rise, in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service