Skip to Content
Merck
All Photos(2)

Key Documents

G0635

Sigma-Aldrich

Guanosine 5′-[β,γ-imido]triphosphate trisodium salt hydrate

≥85% (HPLC), powder

Synonym(s):

β:γ-Imidoguanosine 5′-triphosphate trisodium salt hydrate, 5′-Guanylyl-imidodiphosphate trisodium salt hydrate, GMP-PNP, Gpp(NH)p

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N6Na3O13P3 · xH2O
CAS Number:
Molecular Weight:
588.14 (anhydrous basis)
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Quality Level

Assay

≥85% (HPLC)

form

powder

color

white

solubility

H2O: 50 mg/mL

shipped in

dry ice

storage temp.

−70°C

SMILES string

[Na+].[Na+].[Na+].Nc1nc(O)c2ncn([C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)NP(O)([O-])=O)[C@@H](O)[C@H]3O)c2n1

InChI

1S/C10H17N6O13P3.3Na/c11-10-13-7-4(8(19)14-10)12-2-16(7)9-6(18)5(17)3(28-9)1-27-32(25,26)29-31(23,24)15-30(20,21)22;;;/h2-3,5-6,9,17-18H,1H2,(H,25,26)(H3,11,13,14,19)(H4,15,20,21,22,23,24);;;/q;3*+1/p-3/t3-,5-,6-,9-;;;/m1.../s1

InChI key

QGJNRMUNXAROIT-CYCLDIHTSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Guanosine 5′-[β,γ-imido]triphosphate (GMP-PNP, Gpp(NH)p) is a non-hydrolyzable analog of GTP that binds and irreversibly activates G proteins. Gpp(NH)p is used in a variety of applications that involve GTP binding, including GTP-activation, GTP-inhibition, GTP transport, GTP hydrolysis, and GTP structure stabilization. Since a cycle of GTP binding, hydrolysis, and release is required for the initiation of protein translocation across the endoplasmic reticulum, guanosine 5′-[β,γ-imido]triphosphate is often used in studies of protein synthesis. Gpp(NH)p is used to activate ADP-ribosylation factors and to modulate G proteins involved in cell signaling, protein synthesis and other metabolic processes.

Application

Guanosine 5′-[β,γ-imido]triphosphate trisodium salt hydrate has been used: to study K-RAS interactions with the membrane, for the crystallization of Mycobacterium tuberculosis FtsZ, as a translation initiation inhibitor for the analysis of the 48S pre-initiation complex.
Non-hydrolyzable analog of GTP; the preferred GTP analog to activate ADP-ribosylation factor.

Biochem/physiol Actions

Guanosine 5′-[ß,γ-imido]triphosphate binds and irreversibly activates G proteins. Since a cycle of GTP binding, hydrolysis, and release is required for the initiation of protein translocation across the endoplasmic reticulum, this non-hydrolyzable GTP analog is often used in studies of protein synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Connolly et al.
Science (New York, N.Y.), 252(5009), 1171-1173 (1991-05-24)
The signal recognition particle (SRP) directs signal sequence specific targeting of ribosomes to the rough endoplasmic reticulum. Displacement of the SRP from the signal sequence of a nascent polypeptide is a guanosine triphosphate (GTP)-dependent reaction mediated by the membrane-bound SRP
Laure Yatime et al.
Structure (London, England : 1993), 14(1), 119-128 (2006-01-13)
Eukaryotic and archaeal initiation factors 2 (e/aIF2) are heterotrimeric proteins (alphabetagamma) supplying the small subunit of the ribosome with methionylated initiator tRNA. This study reports the crystallographic structure of an aIF2alphagamma heterodimer from Sulfolobus solfataricus bound to Gpp(NH)p-Mg(2+). aIF2gamma is
J R Hepler et al.
Trends in biochemical sciences, 17(10), 383-387 (1992-10-01)
The family of heterotrimeric guanine nucleotide-binding regulatory proteins (G proteins) serves an essential role in transducing receptor-generated signals across the plasma membrane. Recent findings reveal unexpected functional roles for individual G protein subunits. Thus, GTP-binding alpha-subunits and the beta gamma-subunit
Srinivas Gullapalli et al.
Neuropharmacology, 58(8), 1215-1219 (2010-03-11)
Cannabinoid 1 (CB1) receptors have the ability to change conformation between active (R*) and inactive (R) receptor states. Herein, we further characterize these receptor states using series of saturation radioligand binding studies and their differential displacement binding by various CB1
J D Miller et al.
Ciba Foundation symposium, 176, 147-159 (1993-01-01)
In higher eukaryotes proteins bearing a signal sequence are translocated across the membrane of the endoplasmic reticulum (ER). The initial events of protein translocation are the binding of the signal sequence by the 54 kDa subunit (SRP54) of the signal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service