Skip to Content
Merck
All Photos(4)

Key Documents

FD70S

Sigma-Aldrich

Fluorescein isothiocyanate–dextran

average mol wt 60,000-76,000

Synonym(s):

FITC–Dextran

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

bacterial (Leuconostoc mesenteroides)

conjugate

FITC conjugate

form

powder

mol wt

average mol wt 60,000-76,000

extent of labeling

0.003-0.020 mol FITC per mol glucose

color

yellow to yellow-orange

solubility

H2O: 25 mg/mL, clear to slightly hazy, yellow-orange

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Dextran labeled with fluorescein isothiocyanate for possible use in perfusion studies in animals.
FITC-dextran is used in cardiovascular, microcirculation, perfusion, cell monolayer and cell membrane permeability research as fluorescent flux tracer compound that supports the measurement of processes such as blood flow, membrane damage, vascular drainage and renal elimination. Small FITC-dextrans are also used to measure processes such as endocytosis and cell junction permeability. FITC-dextran 70 kDa (FD-70) may be used as a model compound for development of methods to deliver molecules into protoplasts and to study drug release from structures such as hydrogels and liposomes. FD-70 may be used in microvascular and cell monolayer permeability studies.

Linkage

Similar to FD-70, but produced by Sigma.

Other Notes

To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen-Mei Chen et al.
International journal of molecular sciences, 21(10) (2020-05-18)
Zinc oxide nanoparticles (ZnONPs) are frequently encountered nanomaterials in our daily lives. Despite the benefits of ZnONPs in a variety of applications, many studies have shown potential health hazards of exposure to ZnONPs. We have shown that oropharyngeal aspiration of
Gabriela Hurtado-Alvarado et al.
PloS one, 11(11), e0167236-e0167236 (2016-11-29)
Chronic sleep restriction induces blood-brain barrier disruption and increases pro-inflammatory mediators in rodents. Those inflammatory mediators may modulate the blood-brain barrier and constitute a link between sleep loss and blood-brain barrier physiology. We propose that adenosine action on its A2A
Michael J Shirey et al.
Journal of neurophysiology, 114(1), 309-322 (2015-05-15)
Changes in brain temperature can alter electrical properties of neurons and cause changes in behavior. However, it is not well understood how behaviors, like locomotion, or experimental manipulations, like anesthesia, alter brain temperature. We implanted thermocouples in sensorimotor cortex of
Maira Carrillo et al.
Medicine, 97(39), e12392-e12392 (2018-10-04)
Imaging of placental tissues is a difficult task, because of specific for this organ complex multicellular and 3D tissue structure. The tissue clearing systems (X-CLARITY) system is a valuable tool for the examining the expression of molecular pathways in whole
A Bollinger et al.
Lymphology, 40(2), 52-62 (2007-09-15)
Fluorescence microlymphography (FML) is an almost atraumatic technique used to visualize the superficial skin network of initial lymphatics through the intact skin of man. Visualization was performed with an incident light fluorescence microscope following subepidermal injection of minute amounts of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service