Skip to Content
Merck
All Photos(1)

Key Documents

EMU184491

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Gapdh

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCAAGAAGGTGGTGAAGCAGGCATCTGAGGGCCCACTGAAGGGCATCTTGGGCTACACTGAGGACCAGGTTGTCTCCTGCGACTTCAACAGCAACTCCCACTCTTCCACCTTCGATGCCGGGGCTGGCATTGCTCTCAATGACAACTTTGTCAAGCTCATTTCCTGGTATGACAATGAATACGGCTACAGCAACAGGGTGGTGGACCTCATGGCCTACATGGCCTCCAAGGAGTAAGAAACCCTGGACCACCCACCCCAGCAAGGACACTGAGCAAGAGAGGCCCTATCCCAACTCGGCCCCCAACACTGAGCATCTCCCTCACAATTTCCATCCCAGACCCCCATAATAACAGGAGGGGCCTAGGGAGCCCTCCCTACTCTCTTGAATACCATCAATAAAGTTCGCTGC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feifei Feng et al.
Environmental health perspectives, 120(12), 1692-1698 (2012-09-27)
The role of the Nlrp3 inflammasome in nonallergic airway hyperresponsiveness (AHR) has not previously been reported. Recent evidence supports both interleukin (IL) 1β and short fragments of hyaluronan (HA) as contributors to the biological response to inhaled ozone. Because extracellular
Sureni V Mullegama et al.
European journal of human genetics : EJHG, 23(6), 781-789 (2014-10-02)
Individuals with autism spectrum disorders (ASD) who have an identifiable single-gene neurodevelopmental disorder (NDD), such as fragile X syndrome (FXS, FMR1), Smith-Magenis syndrome (SMS, RAI1), or 2q23.1 deletion syndrome (del 2q23.1, MBD5) share phenotypic features, including a high prevalence of
Seyedmehrad Tavallai et al.
Cancer biology & therapy, 15(5), 578-585 (2014-02-22)
The present studies were to determine whether the multi-kinase inhibitor pazopanib interacted with histone deacetylase inhibitors (HDACI: valproate, vorinostat) to kill sarcoma cells. In multiple sarcoma cell lines, at clinically achievable doses, pazopanib and HDACI interacted in an additive to
Mehrad Tavallai et al.
Journal of cellular physiology, 230(9), 2281-2298 (2015-02-24)
We determined whether the multi-kinase inhibitor sorafenib or its derivative regorafenib interacted with phosphodiesterase 5 (PDE5) inhibitors such as Viagra (sildenafil) to kill tumor cells. PDE5 and PDGFRα/β were over-expressed in liver tumors compared to normal liver tissue. In multiple
Laurence Booth et al.
Molecular cancer therapeutics, 13(10), 2384-2398 (2014-08-12)
The present studies examined the toxic interaction between the non-coxib celecoxib derivative OSU-03012 and phosphodiesterase 5 (PDE5) inhibitors, and also determined the roles of endoplasmic reticulum stress response regulators in cell survival. PDE5 inhibitors interacted in a greater than additive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service