Skip to Content
Merck
All Photos(1)

Key Documents

D9402

Sigma-Aldrich

Dialysis tubing cellulose membrane

avg. flat width 76 mm (3.0 in.)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123100
NACRES:
NB.22

avg. diam.

49 mm , when full

avg. flat width

76 mm (3.0 in.)

capacity

~640 mL/ft

Looking for similar products? Visit Product Comparison Guide

General description

Typical molecular weight cut-off = 14,000.

Packaging

Supplied in rolls, dry

Quality

May contain glycerin and sulfur compounds in more than trace amounts.

Preparation Note

Removal of glycerol included as a humectant can be accomplished by washing the tubing in running water for 3-4 hours. Removal of sulfur compounds can be accomplished by treating the tubing with a 0.3% (w/v) solution of sodium sulfide at 80°C for 1 minute. Wash with hot water (60°C) for 2 minutes, followed by acidification with a 0.2% (v/v) solution of sulfuric acid, then rinse with hot water to remove the acid. This tubing will retain most proteins of molecular weight 12,000 or greater.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asma Manzari-Tavakoli et al.
Scientific reports, 10(1), 22012-22012 (2020-12-18)
The utilization of conductive polymers for fabrication of neural scaffolds have attracted much interest because of providing a microenvironment which can imitate nerve tissues. In this study, polypyrrole (PPy)-alginate (Alg) composites were prepared using different percentages of alginate and pyrrole
Stavroula Nanaki et al.
Polymers, 12(4) (2020-04-11)
In the present study, novel block copolymers of poly(l-lactide)-block-poly(propylene adipate) (PLLA-b-PPAd) were synthesized in two ratios, 90/10 and 75/25 w/w and were further investigated as long-acting injectable (LAI) polymeric matrices in naltrexone base microparticle formulations. The synthesized polymers were characterized
Di Wu et al.
Archives of oral biology, 120, 104930-104930 (2020-10-16)
The aim of this study was to investigate the role of microRNA-152-3p (miR-152-3p) in the osteogenic differentiation of human periodontal ligament stem cells (hPDLSCs). HPDLSCs were isolated and identified using immunofluorescence staining, and their osteogenic differentiation capability was evaluated by
Chiara E Ghezzi et al.
Biomaterials, 35(24), 6236-6247 (2014-05-14)
Airway tracts serve as a conduit of transport in the respiratory system. Architecturally, these are composed of cartilage rings that offer flexibility and prevent collapse during normal breathing. To this end, the successful regeneration of an airway tract requires the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service