D9277
Dialysis tubing cellulose membrane
avg. flat width 10 mm (0.4 in.)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41123100
NACRES:
NB.22
Recommended Products
avg. diam.
6 mm , when full
avg. flat width
10 mm (0.4 in.)
capacity
~10 mL/ft
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Typical molecular weight cut-off = 14,000.
Packaging
Supplied in rolls, dry
Quality
May contain glycerin and sulfur compounds in more than trace amounts.
Preparation Note
Removal of glycerol included as a humectant can be accomplished by washing the tubing in running water for 3-4 hours. Removal of sulfur compounds can be accomplished by treating the tubing with a 0.3% (w/v) solution of sodium sulfide at 80°C for 1 minute. Wash with hot water (60°C) for 2 minutes, followed by acidification with a 0.2% (v/v) solution of sulfuric acid, then rinse with hot water to remove the acid. This tubing will retain most proteins of molecular weight 12,000 or greater.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Zhanna Ignatovich et al.
Molecules (Basel, Switzerland), 26(4) (2021-02-14)
Magnetic nanocomposites based on hydroxyapatite were prepared by a one-step process using the hydrothermal coprecipitation method to sinter iron oxides (Fe3O4 and γ-Fe2O3). The possibility of expanding the proposed technique for the synthesis of magnetic composite with embedded biologically active
Sunil Khatak et al.
Tuberculosis (Edinburgh, Scotland), 125, 102008-102008 (2020-10-16)
The present study aimed to formulate anti-tubercular drugs (Rifampicin, Isoniazid and Pyrazinamide) loaded solid lipid nanoparticles (ATDs-SLNs) using microemulsion technique for oral administration. Central composite designed (CCD) was applied to study the effect of stearic acid (X1), Compritol® 888 ATO
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service