Key Documents
CLS3081250CO
Pyrex® large knob top desiccators
replacement cover for 10.5L large knob top desiccator, 1/cs
Synonym(s):
desiccator covers, desiccator lids
About This Item
UNSPSC Code:
41104510
NACRES:
NB.43
Pricing and availability is not currently available.
material
borosilicate glass
packaging
case of 1
pack of 1
manufacturer/tradename
Corning 3081-250CO
suitability
suitable for (general analytical work)
General description
Note: This is the desiccator cover replacement for the Pyrex desiccator CLS3181250.
The Pyrex large knob top desiccators are designed for general analytical work. The large sturdy knob makes the cover easy to handle. Warning: Not for use with vacuum applications. Do not subject to heat or pressure.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)