Skip to Content
Merck
All Photos(1)

Key Documents

L5178Y-S

93050408, mouse lymph, Lymphoblastoid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

L5178Y-S, 93050408

biological source

mouse lymph

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblastoid

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse DBA/2 Lymphoma

Cell Line Description

Derived from a thymic tumour induced in a DBA/2 mouse by methylcholanthrene. The cells were adapted for in vitro growth. The cells have been widely used in toxicity and radiation studies.

Culture Medium

Fischer′s + 2mM Glutamine + 10% Horse Serum (HS)

Subculture Routine

Maintain cultures between 300,000 to 900,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service