Skip to Content
Merck
All Photos(1)

Documents

HEL92.1.7

92111706, human blood, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

HEL92.1.7, 92111706

biological source

human blood

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian erythroleukaemia

Cell Line Description

Derived from 30 year old male Caucasian. Expresses ß-2 microglobulin and Ia antigen. Used in the study of erythroid cell differentiation and globin gene expression. Analogous to Friend cell leukaemia in mice. K-562 has similar characteristics.

Application

Erythroid cell differentiation studies, induced globin synthesis

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 10
D13S317: 9,11
D16S539: 11
D5S818: 11
D7S820: 7
THO1: 7
TPOX: 11
vWA: 14,17

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. Cell growth can be very slow on resuscitation. Stand the culture flask up overnight using 20% FBS.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service