Jurkat E6.1 Cell Line human
88042803, human blood (leukemic T-cell lymphoblast), Not specified
Synonym(s):
Jurkat Cells, Jurkat Clone E6-1 Cells
About This Item
Recommended Products
product name
Jurkat E6.1 Cell Line human, 88042803, from human blood(leukemic T-cell lymphoblast)
biological source
human blood (leukemic T-cell lymphoblast)
growth mode
Suspension
karyotype
Pseudodiploid, modal No 46
morphology
Not specified
products
Interleukin 2 (IL-2), human alpha interferon
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Cell Line Description
Application
- study cell-independent/vector-free delivery of bovine serum albumin-fluorescein conjugate
- determine the cytotoxicity of the cytolethal distending toxin (CDT) holotoxin and its components
- study the expression of bisphenol A exposed estrogen receptor-β (ERβ) and estrogen-related-receptor-α (ERRα) in vitro
DNA Profile
CSF1PO: 11,12
D13S317: 8,12
D16S539: 11
D5S818: 9
D7S820: 8,10
THO1: 6,9.3
TPOX: 8,10
vWA: 18
Culture Medium
Subculture Routine
Other Notes
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service