Skip to Content
Merck
All Photos(1)

Key Documents

MIA-Pa-Ca-2

85062806, human pancreas, Epithelial

Synonym(s):

MIA PaCa-2 Cells, MIA-PaCa2 Cells, MIAPACA-2 Cells, PaCa2 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

MIA-Pa-Ca-2, 85062806

biological source

human pancreas

growth mode

Adherent

karyotype

Modal no. 61; hypotriploid

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian pancreatic carcinoma

Cell Line Description

An established cell line from an undifferentiated human pancreatic carcinoma. The tumour was taken from a 65 year old Caucasian male. The cells are large with abundant cytoplasm, exhibit a high degree of aneuploidy, have a tendency to grow on top of other cells, eventually growing free in suspension. Sensitive to L-Asparaginase.

Application

MIA-Pa-Ca-2 has been used for the assessment of thyroid receptor transcriptional activity and for α-enolase immunostaining.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10
D13S317: 12,13
D16S539: 10,13
D5S818: 12,13
D7S820: 12,13
THO1: 9,10
TPOX: 9
vWA: 15

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service