Skip to Content
Merck
All Photos(1)

Key Documents

LS180

87021202, human colon, Epithelial-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

LS180, 87021202

biological source

human colon

growth mode

Adherent

karyotype

Modal no. 45 (2n=46)

morphology

Epithelial-like

products

Carcinoembryonic antigen (CEA), Mucin

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian colon adenocarcinoma

Cell Line Description

Derived from a 58 year old female Caucasian with Dukes type B adenocarcinoma of the colon. The cells secrete CEA and mucin and are tumorigenic in nude mice. LS174T (Sigma Catalogue number. 87060401) was derived from the same patient. The cells have not been exposed to trypsin.

Application

Tumourigenicity studies

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 2-5x10,000 cells/cm2 by scraping; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service