Skip to Content
Merck
All Photos(1)

Key Documents

NTERA-2 clone D1 Cell Line human

1071221, human testis, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

NTERA-2 clone D1 Cell Line human, 01071221, caucasian pluripotent embryonal carcinoma

biological source

human testis

growth mode

Adherent

karyotype

Aneuploid

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian pluripotent embryonal carcinoma

Application

Regulation of human embryonic differentiation, neuronal studies

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 10,12
D13S317: 13
D16S539: 11,12,13
D5S818: 9,12
D7S820: 10,12
THO1: 9.3
TPOX: 8
vWA: 18,19

Culture Medium

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:4 i.e. seeding 3-4 x 100,000 cells / cm2; harvest cells by scraping and maintain continuously at high density

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service