Skip to Content
Merck
All Photos(1)

Key Documents

C4926

Sigma-Aldrich

Calmodulin-dependent Protein Kinase II fragment 290-309

≥97% (HPLC)

Synonym(s):

CaM kinase II (290-309)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C103H185N31O24S
CAS Number:
Molecular Weight:
2273.83
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (HPLC)

form

solid

composition

Peptide content, ≥70%

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](C)NC(=O)CNC(=O)[C@H](CCCCN)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](C)NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](Cc1ccccc1)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCCCN)NC(=O)[C@@H](N)CC(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CCSC)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C)C(O)=O

InChI

1S/C103H185N31O24S/c1-17-58(10)80(98(154)131-75(50-57(8)9)97(153)133-82(63(15)136)100(156)134-81(62(14)135)99(155)126-72(39-46-159-16)92(148)127-73(48-55(4)5)93(149)119-61(13)101(157)158)132-84(140)59(11)117-79(138)53-116-86(142)66(33-21-25-40-104)122-95(151)74(49-56(6)7)128-90(146)68(35-23-27-42-106)124-89(145)71(38-30-45-115-103(112)113)125-88(144)70(37-29-44-114-102(110)111)120-83(139)60(12)118-94(150)77(52-78(109)137)130-96(152)76(51-64-31-19-18-20-32-64)129-91(147)69(36-24-28-43-107)123-87(143)67(34-22-26-41-105)121-85(141)65(108)47-54(2)3/h18-20,31-32,54-63,65-77,80-82,135-136H,17,21-30,33-53,104-108H2,1-16H3,(H2,109,137)(H,116,142)(H,117,138)(H,118,150)(H,119,149)(H,120,139)(H,121,141)(H,122,151)(H,123,143)(H,124,145)(H,125,144)(H,126,155)(H,127,148)(H,128,146)(H,129,147)(H,130,152)(H,131,154)(H,132,140)(H,133,153)(H,134,156)(H,157,158)(H4,110,111,114)(H4,112,113,115)/t58-,59-,60-,61-,62+,63+,65-,66-,67-,68-,69-,70-,71-,72-,73-,74-,75-,76-,77-,80-,81-,82-/m0/s1

InChI key

LRKHMNPQPYICFK-RBGVSSEXSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Leu-Lys-Lys-Phe-Asn-Ala-Arg-Arg-Lys-Leu-Lys-Gly-Ala-Ile-Leu-Thr-Thr-Met-Leu-Ala

Biochem/physiol Actions

Potent Ca2+/calmodulin-dependent protein kinase II inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Fukunaga et al.
The Journal of biological chemistry, 268(11), 7863-7867 (1993-04-15)
Among the molecular mechanisms that have been proposed to contribute to long-term potentiation in hippocampus are the activation and autophosphorylation of Ca2+/calmodulin-dependent protein kinase II (CaM kinase II). Here we report that high, but not low frequency stimulation applied to
S Basavappa et al.
Biochemical and biophysical research communications, 254(3), 699-702 (1999-01-28)
Activation of L-type calcium channels in the neuroendocrine, cholecytstokinin-secreting cell line, STC-1, is vital for secretion of CCK. In the present study, the regulation of L-type Ca2+ channels by cAMP and Ca2+ calmodulin dependent protein kinase II (CaM-KII) in STC-1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service