Skip to Content
Merck
All Photos(3)

Key Documents

C4915

Sigma-Aldrich

S-(+)-Chlorpheniramine maleate salt

Synonym(s):

(S)-γ-(4-Chlorophenyl)-N,N-dimethyl-2-pyridinepropanamine maleate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19ClN2 · C4H4O4
CAS Number:
Molecular Weight:
390.86
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

form

powder

Quality Level

originator

Bayer

SMILES string

[H]\C(=C(/[H])C(O)=O)C(O)=O.CN(C)CC[C@@H](c1ccc(Cl)cc1)c2ccccn2

InChI

1S/C16H19ClN2.C4H4O4/c1-19(2)12-10-15(16-5-3-4-11-18-16)13-6-8-14(17)9-7-13;5-3(6)1-2-4(7)8/h3-9,11,15H,10,12H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/t15-;/m0./s1

InChI key

DBAKFASWICGISY-DASCVMRKSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

General description

Chlorpheniramine maleate is a cationic amphiphilic anti-histamine agent, which is composed of a hydrophobic ring structure and a hydrophilic side chain with a charged cationic amino group.[1]

Application

S-(+)-Chlorpheniramine maleate salt has been used to study the anticholinergic effect of Achillea millefolium and Portulaca olerace on muscarinic receptors of guinea pig tracheal smooth muscle.[2][3]

Biochem/physiol Actions

H1 histamine receptor antagonist; active isomer.
Chlorpheniramine maleate is clinically used as a topical ointment to treat skin disorders such as sunburn, urticaria, angioedema, pruritus and insect bites.[1]

Features and Benefits

This compound is featured on the Histamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Bayer. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Sugimoto et al.
Immunopharmacology, 48(1), 1-7 (2000-05-24)
An animal model of chronic allergic rhinitis was developed by repeated local booster sensitization into the nasal cavity in sensitized rats. The severity of allergic rhinitis was assessed by determining the extent of two markers of nasal allergic symptoms (sneezing
Rana Keyhanmanesh et al.
Planta medica, 76(3), 218-222 (2009-08-28)
In previous studies, the relaxant, anticholinergic (functional antagonism) and antihistaminic, effects of Nigella sativa have been demonstrated on guinea pig tracheal chains. In the present study, the prophylactic effect of thymoquinone (one of the constituents of Nigella sativa) on tracheal
T Yoshida et al.
Digestive diseases and sciences, 45(6), 1138-1144 (2000-07-06)
This study aimed to examine the relationship between a harmful effect of histamine and apoptosis following ischemia-reperfusion in the rat intestine. The superior mesenteric artery was occluded for 60 min followed by reperfusion for 60 min. Rats were infused with
Azadeh Feizpour et al.
Indian journal of pharmacology, 45(1), 13-17 (2013-04-02)
To investigate one possible mechanism for the observed relaxant effect of A. millefolium (Achillea millefolium), in the present study the inhibitory effect of the extract of this plant on muscarinic receptors was examined. The effects of three concentrations of aqueous-ethanolic
Cetin Tas et al.
Farmaco (Societa chimica italiana : 1989), 58(8), 605-611 (2003-07-24)
The objective of this study was to evaluate the in vitro and ex vivo percutaneous absorption of chlorpheniramine maleate (CPM) from different hydrogel formulations. Various concentrations of polymers, including hydroxypropylmethylcellulose (HPMC), sodium carboxymethylcellulose (NaCMC) and methyl cellulose (MC) were used

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service