Skip to Content
Merck
All Photos(1)

Key Documents

C1800000

Chlorphenamine maleate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(±)-Chlorpheniramine maleate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19ClN2 · C4H4O4
CAS Number:
Molecular Weight:
390.86
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

chlorpheniramine

manufacturer/tradename

EDQM

mp

130-135 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CN(C)CCC(C1=CC=C(Cl)C=C1)C2=NC=CC=C2.O=C(/C=C\C(O)=O)O

InChI

1S/C16H19ClN2.C4H4O4/c1-19(2)12-10-15(16-5-3-4-11-18-16)13-6-8-14(17)9-7-13;5-3(6)1-2-4(7)8/h3-9,11,15H,10,12H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

DBAKFASWICGISY-BTJKTKAUSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Chlorphenamine maleate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

H1 Histamine receptor antagonist.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shirlei Cristina Moreira et al.
Revista da Sociedade Brasileira de Medicina Tropical, 40(5), 591-593 (2007-11-10)
An occurrence of pruritic papular dermatitis among the whole crew of a Filipino commercial ship in Salvador, State of Bahia, was associated with contact with Hylesia moths. This unusual type of dermatitis is caused by the bristles (flechettes) on the
F Brunet-Possenti et al.
Dermatology (Basel, Switzerland), 223(3), 200-202 (2011-10-12)
Primary cutaneous follicle center lymphoma (PCFCL) is the most common cutaneous B cell lymphoma. It is most often indolent and responds well to rituximab. We present a case of transient rituximab-induced edematous lesions located exclusively on tumor papules in a
E L Theunissen et al.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology, 34(2), 250-258 (2004-02-28)
Mequitazine is a so-called 'non-sedative' second-generation antihistamine even though it has never been firmly established that this drug's sedative potential actually differs from that of the 'sedative' first-generation antihistamines. The present study compares the sedative effects of three doses of
P van Ruitenbeek et al.
Journal of psychopharmacology (Oxford, England), 22(6), 663-672 (2008-01-23)
Results from recent animal studies suggest an important role for histamine in memory functioning. Histaminergic drugs might prove beneficial for people suffering from memory impairment. To determine if histamine is involved in memory functioning this study evaluates the effects of
Ronilson Agnaldo Moreno et al.
Biomedical chromatography : BMC, 24(7), 774-781 (2009-11-20)
In the present study a fast, sensitive and robust validated method to quantify chlorpheniramine in human plasma using brompheniramine as internal standard (IS) is described. The analyte and the IS were extracted from plasma by LLE (diethyl ether-dichloromethane, 80:20, v/v)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service