Skip to Content
Merck
All Photos(1)

Key Documents

C0605

Sigma-Aldrich

Z-Asp(O-Me)-Glu(O-Me)-Val-Asp(O-Me) fluoromethyl ketone

≥90% (HPLC)

Synonym(s):

Z-DEVD-FMK

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H41N4O12F
CAS Number:
Molecular Weight:
668.66
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥90% (HPLC)

form

powder

color

off-white

solubility

DMSO: ≥20 mg/mL, clear

shipped in

dry ice

storage temp.

−20°C

SMILES string

COC(=O)CC[C@H](NC(=O)[C@H](CC(=O)OC)NC(=O)OCc1ccccc1)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC(=O)OC)C(=O)CF

InChI

1S/C30H41FN4O12/c1-17(2)26(29(42)33-20(22(36)15-31)13-24(38)45-4)35-27(40)19(11-12-23(37)44-3)32-28(41)21(14-25(39)46-5)34-30(43)47-16-18-9-7-6-8-10-18/h6-10,17,19-21,26H,11-16H2,1-5H3,(H,32,41)(H,33,42)(H,34,43)(H,35,40)/t19-,20-,21-,26-/m0/s1

InChI key

GBJVAVGBSGRRKN-JYEBCORGSA-N

Biochem/physiol Actions

A cell-permeable inhibitor of caspase 3, 6, 7, 8, 10, which exhibits competitive and irreversible inhibition.

Features and Benefits

This compound is featured on the Caspases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karianne Risberg et al.
PloS one, 6(9), e24012-e24012 (2011-09-15)
In cancer, combinations of drugs targeting different cellular functions is well accepted to improve tumor control. We studied the effects of a Pseudomonas exotoxin A (PE)-based immunotoxin, the 9.2.27PE, and the BH-3 mimetic compound ABT-737 in a panel of melanoma
Eileen Looby et al.
BMC cancer, 9, 190-190 (2009-06-19)
The progression from Barrett's metaplasia to adenocarcinoma is associated with the acquirement of an apoptosis-resistant phenotype. The bile acid deoxycholate (DCA) has been proposed to play an important role in the development of esophageal adenocarcinoma, but the precise molecular mechanisms
Ji-An Zhao et al.
Oncology letters, 12(5), 4252-4262 (2016-11-30)
Curcumin (CUR) has been demonstrated to protect against carcinogenesis and to prevent tumor development in cancer; however, the clinical application of CUR is limited by its instability and poor metabolic properties. The present study offers an strategy for a novel
Minami Goto et al.
Veterinary and comparative oncology, 17(3), 285-297 (2019-02-16)
Tumour necrosis factor-related apoptosis-inducing ligand (TRAIL) is an apoptosis-inducing cytokine that shows potential therapeutic value for human neoplasms, and is effective in some canine tumours; however, its potential for killing canine hemangiosarcoma (HSA) cells is unknown. Thus, we evaluated the
Cagri Bodur et al.
PloS one, 8(2), e56369-e56369 (2013-02-27)
Pramanicin (PMC) is an antifungal agent that was previously demonstrated to exhibit antiangiogenic and anticancer properties in a few in vitro studies. We initially screened a number of PMC analogs for their cytotoxic effects on HCT116 human colon cancer cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service