Skip to Content
Merck
All Photos(1)

Key Documents

B3398

Sigma-Aldrich

Anti-Human IgG2−Biotin antibody, Mouse monoclonal

clone HP-6014, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-Human IgG2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

Quality Level

conjugate

biotin conjugate

antibody form

purified from hybridoma cell culture

antibody product type

secondary antibodies

clone

HP-6014, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

direct ELISA: 1:30,000

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Monoclonal Anti-Human IgG2 (mouse IgG1 isotype) is derived from the hybridoma (HP-60141 ) produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. IgG is the most abundant serum immunoglobulins of the immune system. IgG2 is a subtype of IgG. IgG is secreted by B cells and is found in blood and extracellular fluids.

Specificity

The antibody is specific for human IgG2 and is non-reactive with other IgG subclasses. This clone has been singled out as the most widely applicable IgG2 specific monoclonal antibody by the IUIS/WHO study.

Application

Monoclonal Anti-Human IgG2−Biotin antibody produced in mouse has been used in indirect enzyme linked immunosorbent assay (ELISA) and immunoblot.

Biochem/physiol Actions

IgG provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections.
Low IgG2 levels were found in patients with systemic lupus erythematosus (SLE) and juvenile diabetes melitus.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Immunoglobulin subclass deficiency
Hanson L A, et al.
The Pediatric Infectious Disease Journal, 7, 7-21 (1988)
Nadia Ben Nouir et al.
Journal of clinical microbiology, 46(5), 1631-1640 (2008-03-28)
Echinococcus granulosus protoscolex soluble somatic antigens (PSSAs) were assessed for their prognostic value in the serological follow-up of young patients treated for cystic echinococcosis (CE), compared to conventional hydatid fluid (HF) antigen. Based on different clinical courses and outcome of
Marianna Spatola et al.
Neurology, 95(22), e3012-e3025 (2020-09-16)
To clinically characterize patients with anti-metabotropic glutamate receptor (mGluR) 1 encephalitis, to identify prognostic factors, and to study the immunoglobulin G (IgG) subclasses and effects of antibodies on neuronal mGluR1 clusters. Clinical information on new and previously reported patients was
Esmail Mohamad El-Fakharany et al.
Experimental and therapeutic medicine, 13(4), 1313-1320 (2017-04-18)
Hepatitis C virus (HCV) infection represents a world health problem and no protective vaccine or effective drug currently exists. For economic reasons, many patients use traditional medicines to control the infection. In Egypt, camel milk is one of the traditional
K Akuta et al.
Journal of thrombosis and haemostasis : JTH, 17(1), 206-219 (2018-11-06)
Essentials Acquired Glanzmann thrombasthenia (aGT) is generally caused by function-blocking antibodies (Abs). We demonstrated a unique aGT case due to marked reduction of αIIbβ3 with anti-αIIbβ3 Abs. The anti-αIIbβ3 Abs of the patient did not inhibit platelet function but reduced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service